Skip to Content
Merck
All Photos(1)

Documents

A9285

Sigma-Aldrich

α1-Antichymotrypsin from human plasma

≥92.5% (SDS-PAGE), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human plasma

Quality Level

Assay

≥92.5% (SDS-PAGE)

form

lyophilized powder

mol wt

55-66 kDa

technique(s)

microbiological culture: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... SERPINA3(12)

General description

α1-Antichymotrypsin (α1-ACT) is also known as serpin peptidase inhibitor, clade A member 3 (SERPINA3). It is a 68 kDa serine protease inhibitor, secreted by the liver. It belongs to the serpin superfamily of protease inhibitors. SERPINA3 is located on human chromosome 14q32.1.

Application

α1-Antichymotrypsin from human plasma has been used as a component in yeast extract peptone dextrose broth (YEPD) for biofilm dispersion assays.

Biochem/physiol Actions

α1-Antichymotrypsin (α1-ACT) can block the functions of various serine proteases like chymotrypsin and cathepsin G. It serves as a potential therapeutic target of endometrial carcinoma (EC).
Specific inhibitor of chymotrypsin-like serine proteases. Promotes amyloid-β polymerization in vitro and in vivo. Levels in plasma and cerebrospinal fluid have been found to correlate with progression of dementia in Alzheimer′s patients and induce phosphorylation of tau protein in neurons.

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing Tris buffer salt and NaCl.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is Alpha-1 Antichymotrypsin Gene Polymorphism a Risk Factor for Primary Intracerebral Hemorrhage? A Case-Control Study and Meta-Analysis
Hu X, et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 2149-2149 (2015)
Human plasma proteinase inhibitors.
J Travis et al.
Annual review of biochemistry, 52, 655-709 (1983-01-01)
Up-regulation of SERPINA3 correlates with high mortality of melanoma patients and increased migration and invasion of cancer cells.
Zhou J, et al.
Oncotarget, 8(12), 18712-18712 (2017)
Karin E Darpel et al.
PloS one, 6(3), e17545-e17545 (2011-03-23)
Bluetongue virus (BTV) and epizootic haemorrhagic disease virus (EHDV) are related orbiviruses, transmitted between their ruminant hosts primarily by certain haematophagous midge vectors (Culicoides spp.). The larger of the BTV outer-capsid proteins, 'VP2', can be cleaved by proteases (including trypsin
SERPINA3 promotes endometrial cancer cells growth by regulating G2/M cell cycle checkpoint and apoptosis.
Yang GD
International Journal of Clinical and Experimental Pathology, 7(4), 1348-1358 (2014)

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service