Skip to Content
Merck
All Photos(5)

Documents

T4799

Sigma-Aldrich

Trypsin from porcine pancreas

lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid

Synonym(s):

Cocoonase, Tryptar, Tryptase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75

biological source

Porcine pancreas

Quality Level

product line

BioReagent

form

lyophilized powder

specific activity

1,000-2,000 BAEE units/mg solid

mol wt

23.8 kDa

concentration

25 mg/mL

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

pH

7.6

shipped in

ambient

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Trypsin is applicable for tissue disaggregation, due to its effective action and tolerance towards different cell type and serum-induced neutralization.

Application

For trypsin digestion of peptides, use a ratio of about 1:100 to 1:20 for trypsin:peptide. The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. Trypsins have also been used for the re-suspension of cells during cell culture, in proteomics research for digestion of proteins and in various in-gel digestionsns†. Additional applications include assessing crystallization by membrane-based techniques and in a study to determine that protein folding rates and yields can be limited by the presence of kinetic traps.
Trypsin from porcine pancreas has been used to digest chicken bones. It has also been used in the isolation of luteal endothelial cells.

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Serine protease inhibitors, including DFP, TLCK, APMSF, AEBSEF, and aprotinin, amongst others, will inhibit Trypsin.

Components

Trypsin consists of a single chain polypeptide of 223 amino acid residues, produced by the removal of the N-terminal hexapeptide from trypsinogen which is cleaved at the Lys - lle peptide bond. The sequence of amino acids is cross-linked by 6 disulfide bridges. This is the native form of trypsin, beta-trypsin. BETA-trypsin can be autolyzed, cleaving at the Lys - Ser residue, to produce alpha-trypsin. Trypsin is a member of the serine protease family.

Caution

Solutions in 1 mM HCl are stable for 1 year in aliquots and stored at -20°C. The presence of Ca2+ will also diminish the self-autolysis of trypsin and maintain its stability in solution. Trypsin will also retain most of its activity in 2.0 M urea, 2.0 M guanidine HCl, or 0.1% (w/v) SDS.

Unit Definition

One BAEE unit will produce a ΔA253 of 0.001 per min at pH 7.6 at 25° C using BAEE as substrate. One BTEE unit = 320 ATEE units. Reaction volume = 3.2 mL (1 cm light path).

Preparation Note

This product is a lyophilized powder soluble in Hank′s Balanced Salt Solution at 25 mg/mL.
For applications that require EDTA, solubilizing trypsin should be done with a buffered salt solution contaiing no Ca2+ or Mg2+.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The development of angiotensin I-converting enzyme inhibitor derived from chicken bone protein
Animal Science Journal = Nihon Chikusan Gakkaiho, 79(1), 122-128 (2008)
Prostaglandin F2alpha regulates the nitric oxide generating system in bovine luteal endothelial cells
Lee SH, et al.
Journal of Reproduction and Development, 55(4), 418-424 (2009)
Effects of storage and passage of bovine luteal endothelial cells on endothelin-1 and prostaglandin F2alpha production
Acosta TJ, et al.
Journal of Reproduction and Development, 53(3), 473-480 (2007)
Effects of tea polyphenols on the activities of ?-amylase, pepsin, trypsin and lipase (2007)
Effects of tumor necrosis factor alpha and Interferon gamma on the viability and mRNA expression of TNF receptor type I in endothelial cells from the bovine corpus luteum
Hojo T, et al.
Journal of Reproduction and Development, 56(5), 515-519 (2010)

Protocols

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service