Skip to Content
Merck
All Photos(1)

Documents

C5020

Sigma-Aldrich

Cefazolin sodium salt

89.1-110.1%

Synonym(s):

Cefazoline sodium, Monosodium cefazolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N8NaO4S3
CAS Number:
Molecular Weight:
476.49
Beilstein:
3585038
EC Number:
MDL number:
UNSPSC Code:
51282532
PubChem Substance ID:
NACRES:
NA.76

Quality Level

Assay

89.1-110.1%

form

powder

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)Cn4cnnn4)C3=O)C([O-])=O)s1

InChI

1S/C14H14N8O4S3.Na/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21;/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26);/q;+1/p-1/t9-,12-;/m1./s1

InChI key

FLKYBGKDCCEQQM-WYUVZMMLSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Biochem/physiol Actions

Used to study the effect of expression, binding and inhibition of penicillin-binding proteins, especially PBP2 and PBP4 on bacterial cell wall mucopeptide synthesis.

Preparation Note

Freely soluble in water, very slightly soluble in chloroform and ethanol.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo-Bae Seo et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 106(2), 751-759 (2017-03-24)
An injectable hydrogel system with sustained bone morphogenetic protein 2 (BMP-2) release ability was developed for vertical bone regeneration at peri-implant sites and enhanced osseointegration of dental implants. In three young male beagle dogs, a pair of defects was created
Mei Zhang et al.
Therapeutic drug monitoring, 40(6), 682-692 (2018-07-18)
Pharmacokinetic studies and therapeutic drug monitoring of antibiotics require a simple, rapid, and reliable analytical method for monitoring the concentrations in plasma, including unbound concentrations for highly protein-bound drugs. The aim of the current work was to develop and validate
Francesca D'Este et al.
Journal of peptide science : an official publication of the European Peptide Society, 23(10), 777-789 (2017-07-15)
The prevention of implant-associated infection, one the most feared complications in orthopaedic surgery, remains a major clinical challenge and urges development of effective methods to prevent bacterial colonization of implanted devices. Alpha-helical antimicrobial peptides (AMPs) may be promising candidates in
Keita Kamikura et al.
Journal of materials science. Materials in medicine, 28(9), 132-132 (2017-07-27)
It has been experimentally proven that orally ingested collagen-derived tripeptides (Ctp) are quickly absorbed in the body and effectively promote the regeneration of connective tissues including bone and skin. Ctp are capable to activate osteoblasts and fibroblasts, which eventually promotes
Jabeen Noore et al.
Antimicrobial agents and chemotherapy, 57(3), 1283-1290 (2013-01-01)
The increasing resistance of bacteria to conventional antibiotics and the challenges posed by intracellular bacteria, which may be responsible for chronic and recurrent infections, have driven the need for advanced antimicrobial drugs for effective elimination of both extra- and intracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service