Skip to Content
Merck
All Photos(1)

Key Documents

C4292

Sigma-Aldrich

Cefoperazone sodium salt

870 - 1015 μg/mg anhydrous basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26N9NaO8S2
CAS Number:
Molecular Weight:
667.65
Beilstein:
4902135
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

Assay

870-1015 μg/mg (anhydrous basis)

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SCC(CSc4nnnn4C)=C(N3C2=O)C([O-])=O)c5ccc(O)cc5)C(=O)C1=O

InChI

1S/C25H27N9O8S2.Na/c1-3-32-8-9-33(21(39)20(32)38)24(42)27-15(12-4-6-14(35)7-5-12)18(36)26-16-19(37)34-17(23(40)41)13(10-43-22(16)34)11-44-25-28-29-30-31(25)2;/h4-7,15-16,22,35H,3,8-11H2,1-2H3,(H,26,36)(H,27,42)(H,40,41);/q;+1/p-1/t15-,16-,22-;/m1./s1

InChI key

NCFTXMQPRQZFMZ-WERGMSTESA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Cefoperazone is used to study drug-protein binding, expression and inhibition of penicillin-binding proteins (PBPs) during cell wall synthesis.

Biochem/physiol Actions

Cefoperazone exerts its bactericidal effect by inhibiting the mucopeptide synthesis that affects the structure of bacterial cell wall. It has a dual excretory pattern, primarily via the biliary system and secondarily via the kidney.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Gonik et al.
Antimicrobial agents and chemotherapy, 30(6), 874-876 (1986-12-01)
Limited pharmacokinetic data for cefoperazone are available from the parturient. Because cefoperazone has a dual excretory pattern, primarily via the biliary system and secondarily via the kidney, pregnancy-induced physiologic alterations can influence its deposition and clearance. Twelve term parturients receiving
Mandy Brueckner et al.
Macromolecular bioscience, 20(9), e2000097-e2000097 (2020-07-07)
Conventional therapies for chronic inflammation with high dose application of active agents are often accompanied with severe side effects so that other therapeutical strategies shall be developed to be less physically demanding but still highly efficient. Locally applied Layer-by-Layer (LbL)
Thomas Kaiser et al.
Frontiers in cellular and infection microbiology, 11, 614218-614218 (2021-03-12)
Human microbiota-associated (HMA) mouse models offer a valuable approach to study the role of intestinal microbiota in the development of obesity. In this study, we used an HMA model to evaluate whether engraftment of human obese or lean microbiota, from
Souwelimatou Amadou Amani et al.
Infection and immunity, 88(3) (2019-12-25)
The intracellularly active bacterial toxin TcdB is a major Clostridioides difficile virulence factor that contributes to inflammation and tissue damage during disease. Immunization with an inactive TcdB fragment prevents C. difficile infection (CDI)-associated pathology. The protective immune response against inactive
Andrea T Fessler et al.
Veterinary microbiology, 157(1-2), 226-231 (2011-12-23)
The correct assessment of mastitis pathogens for their susceptibility/resistance to cefoperazone is currently hampered by the lack of harmonized test conditions and interpretive criteria. The aim of this study was to provide a proposal for clinical breakpoints of cefoperazone which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service