Skip to Content
Merck
All Photos(1)

Key Documents

32422

Supelco

Ceftiofur hydrochloride

VETRANAL®, analytical standard

Synonym(s):

(6R,7R)-7-{[(2Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino}-3-{[(2-furanylcarbonyl)thio]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride, 3-[(2-Furylcarbonyl)thio]methyl]-7-[2-(2-amino-4-thiazolyl)-2-(methoxyiminoacetamido)-3-cephem-4-carboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N5O7S3 · HCl
CAS Number:
Molecular Weight:
560.02
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)c3ccco3)=C(N2C1=O)C(O)=O)c4csc(N)n4

InChI

1S/C19H17N5O7S3.ClH/c1-30-23-11(9-7-34-19(20)21-9)14(25)22-12-15(26)24-13(17(27)28)8(5-32-16(12)24)6-33-18(29)10-3-2-4-31-10;/h2-4,7,12,16H,5-6H2,1H3,(H2,20,21)(H,22,25)(H,27,28);1H/b23-11-;/t12-,16-;/m1./s1

InChI key

KEQFDTJEEQKVLM-JUODUXDSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L McLaughlin et al.
Journal of dairy science, 95(8), 4363-4371 (2012-07-24)
The objectives of this study were to evaluate efficacy of a 2-dose regimen of ceftiofur crystalline free acid sterile suspension (CCFA-SS) for treatment of acute metritis in lactating dairy cows under field conditions and to provide additional safety and injection
C L McLaughlin et al.
Theriogenology, 79(4), 725-734 (2013-01-08)
Two studies were conducted to determine if clinical metritis could be prevented or decreased in at-risk lactating dairy cows by a single treatment with Excede Sterile Suspension (ceftiofur crystalline free acid sterile suspension [CCFA-SS]) administered within 24 hours after an
S M Rowe et al.
Journal of dairy science, 103(7), 6473-6492 (2020-05-26)
Selective dry-cow therapy (SDCT) could be used to reduce antibiotic use on commercial dairy farms in the United States but is not yet widely adopted, possibly due to concerns about the potential for negative effects on cow health. The objective
Victoriya V Volkova et al.
PloS one, 7(5), e36738-e36738 (2012-05-23)
Antimicrobial use in food animals may contribute to antimicrobial resistance in bacteria of animals and humans. Commensal bacteria of animal intestine may serve as a reservoir of resistance-genes. To understand the dynamics of plasmid-mediated resistance to cephalosporin ceftiofur in enteric
X von Krueger et al.
Journal of dairy science, 96(2), 1054-1062 (2012-12-25)
Acute puerperal metritis (APM) is one of the most common diseases during the puerperal period. Systemic administration of ceftiofur for 5 consecutive days has been shown to be effective for treatment of APM. The objective of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service