U1128
Uracil
BioReagent, suitable for cell culture
Synonym(s):
2,4(1H,3H)-Pyrimidinedione, 2,4-Dihydroxypyrimidine, 2,4-Pyrimidinediol
About This Item
Recommended Products
biological source
synthetic (organic)
product line
BioReagent
Assay
≥99% (HPLC)
form
powder
technique(s)
cell culture | mammalian: suitable
mp
>300 °C (lit.)
solubility
1 M NaOH: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow
storage temp.
room temp
SMILES string
O=C1NC=CC(=O)N1
InChI
1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)
InChI key
ISAKRJDGNUQOIC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service