Skip to Content
Merck
All Photos(2)

Key Documents

C2399

Sigma-Aldrich

Collagenase from Clostridium histolyticum

0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Quality Level

sterility

0.2 μm filtered

form

lyophilized powder

specific activity

≥4 FALGPA units/mg solid
≥700 CDU/mg solid (CDU = collagen digestion units)

purified by

chromatography

solubility

TESCA buffer (50 mM TES, 0.36 mM Calcium chloride, pH 7.4): soluble 0.05-0.1 mg/mL at 37 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Collagenase from Clostridium histolyticum has been used:
  • to induce intracerebral hemorrhage (ICH) in mice
  • to digest collagen 3D gels for immunofluorescence
  • to cause focal blood extravasation and hemorrhage in mouse

Collagenase has been used in the preparation of arterial tissue for the study of Advanced Glycosylation End Products (AGE). The enzyme has also been used along with other proteases for the disaggregation of human tumor, mouse kidney, human brain, lung epithelium and many other tissues. It is also effective in liver and kidney perfusion studies, digestion of pancreas, and isolation of nonparenchymal hepatocytes.

Biochem/physiol Actions

Clostridium histolyticum contains two classes of collagenases. Clostridial collagenases are more effective than mammalian collagenases. It cleaves at multiple cleavage sites within the collagen triple helix. Clostridial collagenases are involved in bacterial invasion in gas gangrene.
Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
Effective release of cells from tissue requires the action of both collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa (pH optimum: 6.3-8.8). The enzyme recognizes the sequence -R-Pro-8-X-Gly-Pro-R-, where X is most often a neutral amino acid.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Preparation Note

Prepared from Type VII (C0773). A stock solution may be prepared by dissolving 0.05-0.1 mg of collagenase in one mL of 50 mM TES buffer containing 0.36 mM calcium chloride (TESCA), pH 7.4 at 37 °C.

substrate

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The proto-oncoprotein SYT (SS18) controls ATP release and regulates cyst formation by polarized MDCK cells
Chittezhath M, et al.
Experimental Cell Research, 314(19), 3551-3562 (2008)
Silke Proesmans et al.
Frontiers in medicine, 8, 744157-744157 (2021-11-09)
Introduction: [18F]-FDG PET is a widely used imaging modality that visualizes cellular glucose uptake and provides functional information on the metabolic state of different tissues in vivo. Various quantification methods can be used to evaluate glucose metabolism in the brain
Yasunori Asao et al.
Biochemical and biophysical research communications, 529(3), 590-595 (2020-08-02)
Intracerebral hemorrhage (ICH) is one of the most severe subtypes of stroke with high morbidity and mortality. Although a lot of drug discovery studies have been conducted, the drugs with satisfactory therapeutic effects for motor paralysis after ICH have yet
Mohammed Alshareef et al.
International journal of molecular sciences, 23(6) (2022-03-26)
Germinal matrix hemorrhage (GMH) is a devastating disease of infancy that results in intraventricular hemorrhage, post-hemorrhagic hydrocephalus (PHH), periventricular leukomalacia, and neurocognitive deficits. There are no curative treatments and limited surgical options. We developed and characterized a mouse model of
Andrea Comba et al.
Nature communications, 13(1), 3606-3606 (2022-06-25)
Intra-tumoral heterogeneity is a hallmark of glioblastoma that challenges treatment efficacy. However, the mechanisms that set up tumor heterogeneity and tumor cell migration remain poorly understood. Herein, we present a comprehensive spatiotemporal study that aligns distinctive intra-tumoral histopathological structures, oncostreams

Protocols

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service