Skip to Content
Merck
All Photos(3)

Key Documents

C2674

Sigma-Aldrich

Collagenase from Clostridium histolyticum

lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75

biological source

Clostridium histolyticum

Quality Level

type

Type I-A

form

lyophilized powder

specific activity

≥125 CDU/mg solid (CDU = collagen digestion units)
0.5-5.0 FALGPA units/mg solid

mol wt

68-130 kDa

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

pH

6.3-8.8

shipped in

ambient

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Collagenase is a protease which cleaves the triple-helical protein called collagen. There are three types of tissue collagenases, and these belong to the matrix metalloproteinases (MMP) family. Collagenase obtained from Clostridium histolyticum has a very strong activity, as it digests collagen from both ends, at temperatures as low as 4-10°C. Crude collagenase mixtures contain two major enzyme types namely, collagenase and clostripain.

Application

Collagenase from Clostridium histolyticum is used for the following applications:

  • Sertoli cell isolation
  • Used in the comparison of enzymatic methods
  • Used during tissue preparations for immunocytochemistry
  • Testicular sperm extraction
  • Preparation of single cell suspensions
  • Immunofluorescence

This product is suitable for the disaggregation of human tumor, mouse kidney, human adult and fetal brain, lung and many other epithelia tissues. It has also been shown to be effective in liver and kidney perfusion studies, digestion of pancreas, isolation of nonparenchymal rat liver cells and hepatocyte preparation. Collagenase has also been used in the preparation of arterial tissue for the study of Advanced Glycosylation End Products. This enzyme has been tested for the release of heptatocytes at a concentration of approximately 1mg/mL. Concentrations for digestion range from 0.1 to 5mg/mL.

Suitable for use in preparation of single cell suspension for sequencing.

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
The collagenase product is a mixture of enzymes secreted by C. histolyticum, with different products differentiated by the relative ratios of the 10-18 components found in the secreted enzymes. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular materialThe action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.

Caution

As supplied, this product is stable for one year at -20°C. There is no loss in FALGPA or protease activity in 30 days at 37°C, 50°C and -20°C. Solutions of crude collagenase are stable if frozen quickly in aliquots (at 10 mg/mL) and kept frozen at -20°C. Further freeze-thaw cycles will damage the solution. The product retains 100% activity over 7 hours when held on ice.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Preparation Note

This collagenase is obtained from the culture filtrate of Clostridium histolyticum. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. This product is Type I-A. Solutions are typically prepared at 1-2 mg/mL in TESCA buffer (containing 50 mM TES, 0.36 mM Calcium chloride, pH 7.4 at 37°C.

substrate

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evan J Myers et al.
American journal of physiology. Cell physiology, 311(6), C945-C959 (2016-09-30)
The SLC4A11 gene encodes the bicarbonate-transporter-related protein BTR1, which is mutated in syndromes characterized by vision and hearing loss. Signs of these diseases [congenital hereditary endothelial dystrophy (CHED) and Harboyan syndrome] are evident in mouse models of Slc4a11 disruption. However
Nabanita Kundu et al.
Metabolic syndrome and related disorders, 17(6), 314-322 (2019-04-25)
Purpose: We investigated the effect of aerobic and resistance exercise on abdominal subcutaneous fat-derived stromal cells in middle-aged subjects
Yixuan Fang et al.
Aging, 11(14), 4910-4922 (2019-07-23)
Autophagy has been well studied in regulating aging; however, the impact of autophagy in one organ on the aging of other organs has not been documented. In this study, we used a mouse model with deletion of an autophagy-essential gene
J Hanoune et al.
The Journal of biological chemistry, 252(6), 2039-2045 (1977-03-25)
Treatment of rat liver plasma membranes with various commercial preparations of crude collagenase from Clostridium histolyticum at concentrations as low as 1 mug/ml, resulted in activation of the adenylate cyclase system. Maximal activation occurred at 50 to 100 mug/ml of
Joseph R Albe et al.
PLoS pathogens, 15(6), e1007833-e1007833 (2019-06-21)
Rift Valley fever virus (RVFV) causes severe disease in livestock concurrent with zoonotic transmission to humans. A subset of people infected with RVFV develop encephalitis, and significant gaps remain in our knowledge of how RVFV causes pathology in the brain.

Articles

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

See All

Protocols

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service