Skip to Content
Merck
All Photos(1)

Key Documents

COLLH-RO

Roche

Collagenase H

from Clostridium histolyticum

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

Clostridium histolyticum

Quality Level

sterility

non-sterile

form

lyophilized

collagenase activity

0.15 U/mg lyophilizate

packaging

pkg of 100 mg (11074032001)
pkg of 2.5 g (11087789001)
pkg of 500 mg (11074059001)

manufacturer/tradename

Roche

concentration

~1 mg/mL (Isolation of rat hepatocytes)
~2 mg/mL (Isolation of adipocytes)

technique(s)

tissue processing: suitable

color

brown

optimum pH

6.0-8.0

solubility

water: soluble

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

foreign activity

Clostripain 39.798 U/mg
Proteases 14.014 U/mg (Azocoll, based on lyoph)
Trypsin 1.668 U/mg (with BAEE)

storage temp.

2-8°C

General description

Collagenase H is an enzyme mixture prepared from Clostridium histolyticum cultures by filtration, ammonium sulfate precipitation, dialysis, and lyophilization.

Specificity

Collagenase degrades native collagen. Clostripain, trypsin-like enzymes, and neutral proteases also degrade other proteins.

Application

Collagenase from C. histolyticum is used for the dissociation of tissues for the establishment of primary cell cultures. Collagenase H is well suited for the isolation of hepatocytes from rat liver by the collagenase perfusion method. This enzyme preparation is also suitable for the preparation of other types of cells such as endothelial cells from large vessels, and for the isolation of adipocytes from epididymal fat pads of rats.

Physical form

Lyophilizate, nonsterile

Preparation Note

Enzyme activity:
Collagenase activity: >0.15U/mg (according to Wünsch) (+25°C, 4-phenyl-azobenzyl-oxycarbonyl-Pro-Leu-Gly-Pro-D-Arg as the substrate).
Contaminating enzyme activities: trypsin, clostripain, and total proteolytic activity
Collagenase H has a balanced ratio of enzyme activities, and is function tested for the isolation of rat hepatocytes (perfusion method).
Inhibitors:
Collagenase inhibitors: EDTA, EGTA, Cys, His, DTT, 2-mercaptoethanol
Collagenase is not inhibited by serum.
Clostripain inhibitors: TLCK
Trypsin inhibitors: aprotinin, trypsin inhibitor (egg white, soybean)
Activator: Ca2+
Working concentration: 0.5 to 2.5mg/ml,
Approximately 1 mg/ml for the isolation of rat hepatocytes and 2mg/ml for the isolation of adipocytes.
Storage conditions (working solution): -15 to -25°C
The reconstituted solution should be stored at -15 to -25°C for short term storage, -60°C or as described below for long-term storage.

Reconstitution

Reconstitution in any balanced salt solution (e.g., HBSS)

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service