Skip to Content
Merck
All Photos(1)

Key Documents

B3770

Sigma-Aldrich

(+)-Biotinamidohexanoic acid hydrazide

≥90% (TLC), powder

Synonym(s):

(+)-Biotin-ε-aminocaproyl hydrazide, (+)-Biotin-X-hydrazide, (+)-Biotinamidocaproyl hydrazide, BACH, N-(+)-Biotinyl-6-aminohexanoic hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H29N5O3S
CAS Number:
Molecular Weight:
371.50
MDL number:
UNSPSC Code:
12352125
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NC.07

Quality Level

Assay

≥90% (TLC)

form

powder

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)NCCCCCC(=O)NN)[C@@]1([H])NC(=O)N2

InChI

1S/C16H29N5O3S/c17-21-14(23)8-2-1-5-9-18-13(22)7-4-3-6-12-15-11(10-25-12)19-16(24)20-15/h11-12,15H,1-10,17H2,(H,18,22)(H,21,23)(H2,19,20,24)/t11-,12-,15-/m0/s1

InChI key

IJJWOSAXNHWBPR-HUBLWGQQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biotinylation reagent with aminocaproic spacer for aldehyde groups (e.g., periodate-oxidized sugars) and carboxylic acids. Typically used for coupling to glycoproteins through the carbohydrate by hydrazone bond formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Spiegel et al.
Journal of immunology (Baltimore, Md. : 1950), 127(2), 572-575 (1981-08-01)
Biotin hydrazide was attached covalently to the aldehyde groups produced by periodate oxidation of bovine brain gangliosides. These modified gangliosides were incorporated into mature rat thymocytes by incubation of the biotinyl gangliosides in the culture medium containing these cells. Avidin
Kimberly MacKay et al.
Briefings in functional genomics, 19(4), 292-308 (2020-05-01)
The advent of high-resolution chromosome conformation capture assays (such as 5C, Hi-C and Pore-C) has allowed for unprecedented sequence-level investigations into the structure-function relationship of the genome. In order to comprehensively understand this relationship, computational tools are required that utilize
Nobuaki Higashi et al.
Biochemical and biophysical research communications, 520(1), 152-158 (2019-10-05)
We examined whether chondroitin sulfates (CSs) exert inhibitory effects on heparanase (Hpse), the sole endoglycosidase that cleaves heparan sulfate (HS) and heparin, which also stimulates chemokine production. Hpse-mediated degradation of HS was suppressed in the presence of glycosaminoglycans derived from
M B Rosenberg et al.
Journal of neurochemistry, 46(2), 641-648 (1986-02-01)
beta-nerve growth factor (NGF) was modified by biotinylation via carboxyl group substitution (C-bio-NGF) using biotin hydrazide and the coupling reagent 1-ethyl-3-(3-dimethylaminopropyl)-carbodiimide, under reaction conditions that yielded an average of 3 biotin additions per NGF subunit. NGF was also biotinylated through
Adelaide U P Hain et al.
Journal of medicinal chemistry, 57(11), 4521-4531 (2014-05-03)
Atg8 is a ubiquitin-like autophagy protein in eukaryotes that is covalently attached (lipidated) to the elongating autophagosomal membrane. Autophagy is increasingly appreciated as a target in diverse diseases from cancer to eukaryotic parasitic infections. Some of the autophagy machinery is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service