Skip to Content
Merck
All Photos(2)

Key Documents

B5161

Sigma-Aldrich

Biotin 3-sulfo-N-hydroxysuccinimide ester sodium salt

≥90% (TLC), powder

Synonym(s):

Sulfosuccinimidyl biotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N3NaO8S2
CAS Number:
Molecular Weight:
443.43
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.46

Assay

≥90% (TLC)

form

powder

solubility

H2O: 10 mg/mL
DMF: 50 mg/mL

storage temp.

−20°C

SMILES string

[Na].OS(=O)(=O)C1CC(=O)N(OC(=O)CCCCC2SCC3NC(=O)NC23)C1=O

InChI

1S/C14H19N3O8S2.Na.H/c18-10-5-9(27(22,23)24)13(20)17(10)25-11(19)4-2-1-3-8-12-7(6-26-8)15-14(21)16-12;;/h7-9,12H,1-6H2,(H2,15,16,21)(H,22,23,24);;

InChI key

NKHCQKKDLWGBRK-UHFFFAOYSA-N

General description

Biotin:avidin second antibody signal systems involve the conjugation of biotin to a secondary antibody. Once the secondary antibody binds to a target primary antibody in an immunochemical assay, the signal is developed by allowing avidin which has been conjugated to an enzyme or signal molecule to bind to the antibody associated biotin molecule.

Application

Water soluble biotinylation reagent. Typically coupled to primary amine in the pH range 6.5-8.5.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Bor Tsai et al.
Biotechnology and bioengineering, 98(2), 498-507 (2007-03-28)
Cell adhesion to a scaffold is a prerequisite for tissue engineering. Many studies have been focused on enhancing cell adhesion to synthetic materials that are used for scaffold fabrication. Previously, we showed that immobilization of biotin molecules to chondrocyte surfaces
Wei-Bor Tsai et al.
Biomaterials, 26(16), 3141-3151 (2004-12-18)
Cell adhesion to synthetic biomaterials is a prerequisite for anchorage cell culture and tissue engineering. The current study investigated utilization of an avidin-biotin binding system in enhancing chondrocyte adhesion to tissue culture polystyrene (TCPS). Biotinylated chondrocytes adhered to avidin-coated TCPS
P Pantano et al.
Analytical chemistry, 65(5), 623-630 (1993-03-01)
Dehydrogenase enzymes are immobilized onto carbon-fiber microelectrode surfaces via avidin-biotin technology and a covalently linked hydrophilic tether. The avidin-biotin coupling strategy allows the selectivity of the electrochemical measurement to be easily changed without reoptimizing the immobilization conditions. Optimized derivatization conditions
Dominic Kamps et al.
Cell reports, 33(9), 108467-108467 (2020-12-03)
Local cell contraction pulses play important roles in tissue and cell morphogenesis. Here, we improve a chemo-optogenetic approach and apply it to investigate the signal network that generates these pulses. We use these measurements to derive and parameterize a system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service