Skip to Content
Merck
All Photos(1)

Key Documents

B0631

Sigma-Aldrich

5-Bromo-2′-deoxyuridine 5′-triphosphate sodium salt

≥90%

Synonym(s):

5-BrdUTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14BrN2O14P3
CAS Number:
Molecular Weight:
547.04
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥90%

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Na].OC1CC(OC1COP(O)(=O)OP(O)(=O)OP(O)(O)=O)N2C=C(Br)C(=O)NC2=O

InChI

1S/C9H14BrN2O14P3.Na.H/c10-4-2-12(9(15)11-8(4)14)7-1-5(13)6(24-7)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h2,5-7,13H,1,3H2,(H,19,20)(H,21,22)(H,11,14,15)(H2,16,17,18);;

InChI key

UWZGQOWZUSBZMC-UHFFFAOYSA-N

General description

5-Bromo-2′-deoxyuridine 5′-triphosphate is used for incorporation into DNA for studying normal and tumor cell proliferation profiles.

Application

5-Bromo-2′-deoxyuridine 5′-triphosphate (5-BrdUTP) may be used as a labeling TdT substrate for terminal deoxynucleotidyl transferase (TdT) reactions or for DNA synthesis. BrdUTP is also used as an effective mutagenic agent. 5-BrdUTP labeling may be detected by immunological techniques or 5-BrdUTP may be derivitized by biotinylation or other methods for detection by fluorogenic or chromogenic methods.
5-Bromo-2′-deoxyuridine 5′-triphosphate sodium salt has been used:
  • in the labelling of 3′-OH termini of fragmented DNA with double stranded breaks in african green monkey kidney cells
  • to label cleaved DNA ends in apoptosis assay in avian tissue sections
  • for incorporation into mice prostates for immunohistochemistry studies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Visualization of heavy ion tracks by labeling 3'-OH termini of induced DNA strand breaks
Konishi T, et al.
Journal of Radiation Research, 52(4), 433-440 (2011)
Effect of 5-fluoro-2?-deoxyuridine (FdUrd) on 5-bromo-2?-deoxyuridine (BrdUrd) incorporation into DNA, measured with a monoclonal BrdUrd antibody and by the BrdUrd/hoechst quenching effect
Ellwart J and Dormer P
Cytometry, 6(6), 513-520 (1985)
Xiangdong Ma et al.
Molecular biology reports, 35(4), 663-667 (2007-09-18)
To explore the mutagenic properties of the nucleotide analogue bromodeoxyuridine triphosphate (BrdUTP), the wild type alpha-amylase (xamy) gene from Xanthomonas campestris pv. campestris 8004 was used as a mutational target. It was mutated using PCR techniques to partially replace deoxythymidine
Zbigniew Darzynkiewicz et al.
Methods in molecular biology (Clifton, N.J.), 682, 91-101 (2010-11-09)
Extensive DNA fragmentation that generates a multitude of DNA double-strand breaks (DSBs) is a hallmark of apoptosis. A widely used approach to identify apoptotic cells relies on labeling DSBs in situ with fluorochromes. Flow or image cytometry is then used
Eisuke Gotoh
Chromosoma, 116(5), 453-462 (2007-05-16)
A basic question of cell biology is how DNA folds to chromosome. Numbers of examples have suggested the involvement of DNA replication in chromosome structure formation. To visualize and identify the dynamics of chromosome structure formation and to elucidate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service