Skip to Content
Merck
All Photos(1)

Key Documents

A9627

Sigma-Aldrich

Ala-Ala-Ala

≥98% (TLC)

Synonym(s):

Tri-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17N3O4
CAS Number:
Molecular Weight:
231.25
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Ala-Ala-Ala,

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

C[C@H](N)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C9H17N3O4/c1-4(10)7(13)11-5(2)8(14)12-6(3)9(15)16/h4-6H,10H2,1-3H3,(H,11,13)(H,12,14)(H,15,16)/t4-,5-,6-/m0/s1

InChI key

BYXHQQCXAJARLQ-ZLUOBGJFSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Ala-Ala

Application

Trialanine (Ala-Ala-Ala) may be used along with other short chain alanines, tetra- and penta-alanine, as model compounds to study physicochemical parameters of small peptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fatma Eker et al.
Journal of the American Chemical Society, 125(27), 8178-8185 (2003-07-03)
Determination of the precise solution structure of peptides is of utmost importance to the understanding of protein folding and peptide drugs. Herein, we have measured the UV circular dichroism (UVCD) spectra of tri-alanine dissolved in D(2)O, H(2)O, and glycerol. The
Kaicong Cai et al.
Physical chemistry chemical physics : PCCP, 11(40), 9149-9159 (2009-10-09)
A molecular mechanics (MM) force field-based empirical electrostatic potential map (MM map) for amide-I vibrations is developed with the aim of seeking a quick and reasonable approach to computing local mode parameters and their distributions in solution phase. Using N-methylacetamide
Róisín Moriarty et al.
Thrombosis and haemostasis, 113(2), 290-304 (2014-11-22)
The integrin αIIbβ3 on resting platelets can bind to immobilised fibrinogen resulting in platelet spreading and activation but requires activation to bind to soluble fibrinogen. αIIbβ3 is known to interact with the general integrin-recognition motif RGD (arginine-glycine-aspartate) as well as
Reinhard Schweitzer-Stenner
The journal of physical chemistry. B, 113(9), 2922-2932 (2009-02-27)
The conformational preference of individual amino acid residues in the unfolded state of peptides and proteins is the subject of a continuous debate. Research has mostly been focused on alanine, owing to its abundance in proteins and its relevance for
Wenfei Li et al.
The Journal of chemical physics, 130(21), 214108-214108 (2009-06-11)
Biomolecular systems are inherently hierarchic and many simulation methods that try to integrate atomistic and coarse-grained (CG) models have been proposed, which are called multiscale simulations. Here, we propose a new multiscale molecular dynamics simulation method which can achieve high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service