Skip to Content
Merck
All Photos(1)

Documents

A5545

Sigma-Aldrich

Anti-Guinea Pig IgG (whole molecule)−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Guinea pig IgG (whole molecule)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

guinea pig

technique(s)

direct ELISA: 1:50,000-1:80,000
dot blot: 1:50,000-1:100,000 (chemiluminescent)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400-1:800

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

An immunoglobulin has two heavy chains and two light chains connected by disulfide bond. It is a glycoprotein. IgG is a major class of immunoglobulin. It has two subclasses of IgG- IgG1 and IgG2.

Specificity

Rabbit anti-guinea pig IgG (whole molecule)-peroxidase antibody binds to guinea pig IgG.

Immunogen

guinea pig IgG

Application

Anti-Guinea Pig IgG (whole molecule) Peroxidase antibody produced in rabbit has been used in diphtheria serology enzyme-linked immunosorbent assay (ELISA) and immunoblotting.

Biochem/physiol Actions

IgGs are glycoprotein antibodies that modulate several immune responses. Guinea pig IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-guinea pig IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins.
Immunoglobulin G (IgG) is the major class and most abundant immunoglobulin of immune system released by plasma cells. IgG mediate immune response with respect to the type of antigen and is found in blood or extracellular fluid. IgG defends against bacterial and viral infections. Maternal IgG crosses from mother to fetus through the placental barrier by pinocytosis to provide passive immunity for the neonate and protect against infections.
Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. IgG1 helps in complement fixation through alternate pathway and IgG2 fixes complement by conventional mechanism. It mainly helps in immune defense. Guinea Pig is a highly preferred laboratory animal to study the local and anaphylactic reactions.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Storage and Stability

Store at -20 °C for long term use. For continuous use, the product may be stored at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots at -20 °C. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig J Bierle et al.
PloS one, 12(11), e0187720-e0187720 (2017-11-04)
Primary Zika virus (ZIKV) infections that occur during pregnancy can cause spontaneous abortion and profoundly disrupt fetal development. While the full range of developmental abnormalities associated with congenital Zika syndrome is not yet known, severe cases of the syndrome can
A Gram et al.
Reproduction in domestic animals = Zuchthygiene, 49 Suppl 2, 41-49 (2014-06-21)
Oxytocin (OT) plays an important role as an inducer of uterine contractility, acting together with its receptor (OTR) to increase synthesis of prostaglandins. Although OT is commonly used in the treatment for dystocia and uterine inertia in the bitch, little
Serotonin transporter inhibition and 5-HT2C receptor activation drive loss of cocaine-induced locomotor activation in DAT Val559 mice
Stewart A, et al.
Neuropsychopharmacology, 44(5), 994-1006 (2018)
Placental structure, function and drug transfer
Griffiths SK and Campbell JP
Encyclopedia of Chemical Processing and Design, 15(2), 84-89 (2014)
Aldo Seguel et al.
Plant physiology, 176(3), 2515-2531 (2018-02-14)
Salicylic acid (SA) is a major defense signal in plants. In Arabidopsis (Arabidopsis thaliana), the chloroplast-localized isochorismate pathway is the main source of SA biosynthesis during abiotic stress or pathogen infections. In the first step of the pathway, the enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service