Skip to Content
Merck
All Photos(1)

Key Documents

AAS18

Supelco

Amino Acid Standard

Amino Acid Standard

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
eCl@ss:
32160406
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

storage temp.

2-8°C

Application

This analytical standard was used as follows:
  • Determination of L- and D-amino acids and glycine by liquid chromatography-tandem mass spectrometry (LC-MS/MS) following their derivatization with chiral Marfey’s reagent
  • High-performance liquid chromatography (HPLC) based amino acid analysis during the characterization of defatted rice bran (DRB) protein concentrate to evaluate its iron binding ability
  • Development of a gas chromatography-mass spectrometry (GC-MS) procedure for the profiling of 112 amino-carboxylic metabolites in human urine samples using 1,1,1,2,2,3,3-heptafluorobutyl chloroformate for derivatization and liquid-liquid microextraction for sample treatment
  • Non-enantiomeric detection and quantification of amino acids in their various forms— free, dissolved, particulate, and total, in natural water samples by HPLC coupled with fluorescence detection using a C18 column

Packaging

1 mL size packaged in flame sealed ampules.

Components

The following components are included in the amino acid standard:
  • L-Alanine
  • Ammonium chloride
  • L-Arginine
  • L-Aspartic acid
  • L-Cystine
  • L-Glutamic acid
  • Glycine
  • L-Histidine
  • L-Isoleucine
  • L-Leucine
  • L-Lysine
  • L-Methionine
  • L-Phenylalanine
  • L-Proline
  • L-Serine
  • L-Threonine
  • L-Tyrosine
  • L-Valine

Other Notes

Amino acids in this standard are 2.5 μmoles per mL in 0.1 N HCl, except L-cystine at 1.25 μmoles per mL.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manish Kumar Patel et al.
BMC plant biology, 20(1), 116-116 (2020-03-17)
Developing fruit is considered as an excellent model to study the complex network of metabolites which are altered rapidly during development. Metabolomics revealed that developing psyllium fruit is a rich source of primary metabolites (ω-3 and ω-6 fatty acids and
Ashok Panda et al.
Plant physiology and biochemistry : PPB, 158, 284-296 (2020-11-27)
Haloxylon salicornicum is a xero-halophyte growing in saline and arid regions of the world. Metabolite profiling was carried out in shoot of both control and salinity treated (400 mM NaCl) samples by GC-QTOF-MS and HPLC-DAD analysis to decipher the salinity tolerance
Benyamin Khoshnevisan et al.
Bioresource technology, 290, 121743-121743 (2019-07-20)
Resource efficient and novel practices to produce proteinaceous food and feed sources can partially alleviate the protein scarcity problem. The conversion of low-value waste streams into single cell protein (SCP) seems a potent solution. This study evaluated the possibility of
Lu Meng et al.
Microbial cell factories, 19(1), 134-134 (2020-06-24)
The Saccharomyces cerevisiae Snf1 complex is a member of the AMP-activated protein kinase family and plays an important role in response to environmental stress. The α catalytic subunit Snf1 regulates the activity of the protein kinase, while the β regulatory
Jean Farup et al.
SpringerPlus, 5, 382-382 (2016-04-12)
Whey protein is generally found to be faster digested and to promote faster and higher increases in plasma amino acid concentrations during the immediate ~60 min following protein ingestion compared to casein. The aim of the present study was to compare

Reviews

Active Filters

  1. France
    • Review 1
    • Vote 1
    5 out of 5 stars.

    great product

    great product that is provided here - fit for purpose

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service