Skip to Content
Merck
All Photos(5)

Key Documents

A0132

Sigma-Aldrich

Atropine

≥99% (TLC), powder

Synonym(s):

Hyoscyamine, Tropine tropate, endo-(±)-α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO3
CAS Number:
Molecular Weight:
289.37
Beilstein:
91260
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (TLC)

form

powder

color

white

solubility

H2O: 2 mg/mL
dilute aqueous acid: soluble
ethanol: soluble

SMILES string

CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16?

InChI key

RKUNBYITZUJHSG-SPUOUPEWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atropine, a belladonna alkaloid, is an anti-muscarinic drug. It is extracted from Atropa belladonna. Atropine is an anticholinergic agent, which is linked to passive mydriasis. It blocks acetylcholine receptors in brain and has hallucinogenic effects. Atropine eye drops are used to treat uveitis and early amblyopia. It is also used to treat nerve agents.

Application

Atropine has been used to study it′s in vitro cytotoxicity, antiviral and antimicrobial activities. It has also used to antagonize acetylcholine induced K+ current.
Atropine has been used:
  • as a tastant to analyse the bitter taste perception in Drosophila larvae
  • as a muscarinic acetylcholine autoreceptors (mAChRs) antagonist to investigate the involvement of individual mAChR subtypes in the control of synapse elimination
  • to investigate the pathways of serotonin (5-HT) action in zebrafish hearts

Biochem/physiol Actions

Atropine is used to treat myopia and ocular axial elongation in Asian children. Small dosage of atropine is used to slow down the heart rate.
Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Reconstitution

Solutions stable for several days at 4 °C.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atropine for the treatment of childhood myopia
Chua WA, et al.
Ophthalmology, 113(12), 2285-2291 (2006)
Weapons of Mass Destruction: An Encyclopedia of Worldwide Policy, Technology, and History, Volume 2 (2005)
Presynaptic muscarinic acetylcholine autoreceptors (M 1, M 2 and M 4 subtypes), adenosine receptors (A 1 and A 2A) and tropomyosin-related kinase B receptor (TrkB) modulate the developmental synapse elimination process at the neuromuscular junction
Nadal L, et al.
Molecular Brain, 9(1), 67-67 (2016)
Muscarinic receptor hyperpolarizes cochlear hair cells of chick by activating Ca (2+)-activated K+ channels.
Shigemoto T, et al.
The Journal of Physiology, 442(1), 669-690 (1991)
Tarascon Pharmacopoeia 2014 Professional Desk Reference Edition (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service