Skip to Content
Merck
All Photos(1)

Key Documents

PHR1379

Supelco

Atropine Sulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Atropine sulfate salt monohydrate, α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Tropine tropate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H23NO3)2 · H2SO4 · H2O
CAS Number:
Molecular Weight:
694.83
Beilstein:
6109275
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 23
traceable to Ph. Eur. A1400000
traceable to USP 1045009

API family

atropine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

189-192 °C (A) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.OS(O)(=O)=O.CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3.CN4[C@H]5CC[C@@H]4C[C@@H](C5)OC(=O)C(CO)c6ccccc6

InChI

1S/2C17H23NO3.H2O4S.H2O/c2*1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12;1-5(2,3)4;/h2*2-6,13-16,19H,7-11H2,1H3;(H2,1,2,3,4);1H2/t2*13-,14+,15+,16?;;

InChI key

JPKKQJKQTPNWTR-CHYDPLAESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Atropine Sulfate is an alkaloid extracted from belladonna herb. It is a cholinergic drug, administered as an anti-arrhythmic medicine.

Application

Atropine Sulfate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by electrophoresis and chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC4087 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of atropine in pharmaceutical preparations by liquid chromatography with fluorescence detection.
Takahashi M, et al.
Journal of Chromatography A, 775(1-2), 137-141 (1997)
Determination of atropine sulfate using a novel sensitive DNA-biosensor based on its interaction on a modified pencil graphite electrode.
Ensafi AA, et al.
Talanta, 131, 149-155 (2015)
LC determination of atropine sulfate and scopolamine hydrobromide in pharmaceuticals.
Ceyhan T, et al.
Journal of Pharmaceutical and Biomedical Analysis, 25(3-4), 399-406 (2001)
Development and validation of a capillary zone electrophoresis method for the determination of atropine, homatropine and scopolamine in ophthalmic solutions.
Cherkaoui S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 696(2), 283-290 (1997)
Miklós Palotai et al.
Neurochemical research, 39(8), 1477-1483 (2014-06-05)
The nociceptin/orphanin FQ peptide (NOP) receptor and its endogenous ligand plays role in several physiologic functions of the central nervous system, including pain, locomotion, anxiety and depression, reward and drug addiction, learning and memory. Previous studies demonstrated that the NOP-receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service