Skip to Content
Merck
All Photos(2)

Key Documents

1.06469

Supelco

Sodium hydroxide

pellets for analysis (max. 0.02% K) EMSURE® ACS,Reag. Ph Eur,ISO

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
EC Index Number:
215-185-5
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

EMSURE®

Assay

99-100% (total alkilinity calculated as NaOH)

form

pellets

impurities

≤0.0003% Total nitrogen (N)

pH

>14 (20 °C, 100 g/L in H2O)

mp

318 °C (lit.)

solubility

water: 1090 g/L at 20 °C

density

2.13 g/cm3 at 20 °C

anion traces

carbonate (as Na2CO3): ≤0.5%
chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.0005%
silicate (as SiO2): ≤0.001%
sulfate (SO42-): ≤0.0005%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ca: ≤0.0005%
Cu: ≤0.0002%
Fe: ≤0.0005%
Hg: ≤0.00001%
K: ≤0.020%
Mg: ≤0.0005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Zn: ≤0.001%
heavy metals (as Ag): ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Biosorbent derived from coffee husk for efficient removal of toxic heavy metals from wastewater.: Researchers developed a biosorbent from coffee husk to remove heavy metals from wastewater. Sodium hydroxide plays a critical role in the chemical activation of the coffee husk, enhancing its adsorption capacity (Quyen et al., 2021).

  • Development of Advanced Chemometric-Assisted Spectrophotometric Methods for the Determination of Cromolyn Sodium and Its Alkaline Degradation Products.: This paper presents advanced spectrophotometric methods for analyzing cromolyn sodium and its degradation products. Sodium hydroxide is used to induce the degradation of cromolyn sodium, allowing for the study of its breakdown products (El Zahar et al., 2020).

  • High resolution and high throughput analytical methods for d-tagatose and process related impurities using capillary electrophoresis.: The study develops capillary electrophoresis methods for analyzing d-tagatose and its impurities. Sodium hydroxide is employed as an electrolyte to enhance the separation and resolution in the electrophoresis process (Surapureddi et al., 2020).

  • Phosphorus in recycling fertilizers - analytical challenges.: This research addresses the analytical challenges in determining phosphorus in recycling fertilizers. Sodium hydroxide is used to adjust the pH of samples, which is crucial for accurate phosphorus quantification (Krüger and Adam, 2017).

Linkage

Replaces: SX0590; SX0590-20

Analysis Note

Assay (acidimetric,NaOH): 99 - 100 %
Assay (total alkalinity calc. as NaOH): 99 - 100 %
Identity: passes test
Appearance of solution: passes test
Carbonate (as Na₂CO₃): ≤ 0.5 %
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.0005 %
Silicate (SiO₂): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.0003 %
Heavy metals (as Ag): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.0005 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
Hg (Mercury): ≤ 0.00001 %
K (Potassium): ≤ 0.020 %
Mg (Magnesium): ≤ 0.0005 %
Ni (Nickel): ≤ 0.0005 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service