Skip to Content
Merck
All Photos(1)

Key Documents

70215

Sigma-Aldrich

2,3-Naphthalenedicarboxaldehyde

suitable for fluorescence

Synonym(s):

NDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CHO)2
CAS Number:
Molecular Weight:
184.19
Beilstein:
2207267
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

form

crystals

Quality Level

impurities

≤2% mono- and dicarboxylic acid (1H-NMR)

mp

131-133 °C (lit.)
132-135 °C

fluorescence

λex 420 nm; λem ~480 nm in 0.1 M borate pH 9.3 (after derivatization with glycine [~90 μM glycine, ~20 μM N-])

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cc2ccccc2cc1C([H])=O

InChI

1S/C12H8O2/c13-7-11-5-9-3-1-2-4-10(9)6-12(11)8-14/h1-8H

InChI key

ZIPLKLQPLOWLTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Naphthalenedicarboxaldehyde, also known as NDA, is a novel fluorescent derivatization agent. The fluorescence max. excitation of NDA is in the range of 424 nm and max. emission is achieved at 484nm.[1]

2,3-Naphthalenedicarboxaldehyde is a fluorescent derivatization agent of primary amines, amino acids, and small peptides. The reaction between the amino compounds and NDA results in highly fluorescent and stable derivative compounds.

Application

2,3-Naphthalenedicarboxaldehyde is used as a fluorescent labelling agent for quantification of Histamine in human immunoglobulin preparations.[1] NAD is used as a derivatization agent for glutathione to quantify protein glutathionylation.[2] It is a suitable reagent used for the development of fluorescence-based microtiter plate assay for γ-glutamylcyclotransferase.[3]

Features and Benefits

NDA based fluorescent assays have the following benefits over O-Phthalaldehyde (OPA):

  • High Reaction rate.
  • High Fluorescence quantum yield.
  • No uncommon excitation wavelength.
  • No side reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Hui et al.
Electrophoresis, 33(12), 1911-1920 (2012-06-29)
Asymmetric dimethylarginine (ADMA) is an inhibitor of nitric oxide synthase and a risk factor for cardiovascular events. We have developed a new derivatization method to enable baseline separation of the regio-isomers, ADMA, and symmetric dimethylarginine (SDMA), within 15 min on
S.M. Lunte et al.
LC-GC, 7, 908-908 (1989)
Jung-Hwan Kim et al.
Osong public health and research perspectives, 2(2), 127-134 (2011-09-01)
To develop and optimize quantitative HPLC method using 2,3-naphthalenedicarboxaldehyde (NDA) after simple and efficient solid phase extraction to determine the histamine in a biopharmaceutical (Histobulin™). The HPLC method was established using NDA-induced Histobulin and compared with the recently reported HPLC
P. de Montigny et al.
Analytical Chemistry, 59, 1096-1096 (1987)
An improved mechanism-based cross-linker for multiplexed kinase detection and inhibition in a complex proteome.
Kai Liu et al.
Chembiochem : a European journal of chemical biology, 9(12), 1883-1888 (2008-07-16)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service