345571
1,5,7-Triazabicyclo[4.4.0]dec-5-ene
98%
Synonym(s):
1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, Hhpp, TBD
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H13N3
CAS Number:
Molecular Weight:
139.20
Beilstein:
3242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
crystals
mp
125-130 °C (lit.)
solubility
acetonitrile: soluble
ethanol: soluble
organic solvents: soluble
water: soluble
SMILES string
C1CNC2=NCCCN2C1
InChI
1S/C7H13N3/c1-3-8-7-9-4-2-6-10(7)5-1/h1-6H2,(H,8,9)
InChI key
FVKFHMNJTHKMRX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
1,5,7-Triazabicyclo[4.4.0]dec-5-ene, a bicyclic guanidine base, has been found to be an excellent catalyst for Michael and Michael-type reactions. It forms 1:1 complex with lasalocid acid and crystal structure of the complex has been studied by X-ray diffraction, FT-IR spectroscopy and 1H NMR.
Application
1,5,7-Triazabicyclo[4.4.0]dec-5-ene may be used as organocatalyst for aminolysis of esters. It may be used as catalyst for direct addition of P(O)-H bonds (dialkyl phosphites and diphenyl phosphonite) across various activated alkenes. Polymer supported 1,5,7-triazabicyclo[4.4.0]dec-5-ene (PTBD) was used as a base and a reagent scavenger for the synthesis of aryl ethers from phenols and alkyl or aryl halides.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jinfeng Wang et al.
Journal of surgical oncology, 111(8), 992-999 (2015-05-16)
We aimed to investigate potential of urinary cell-free microRNA-214 (miR-214) as a noninvasive biomarker for bladder cancer in this report. We screened miR-214 expression in medium from 2 bladder cancer cell lines to determine whether it is secretory. Then we
1, 5, 7-Triazabicyclo [4.4. 0] dec-5-ene (TBD) catalyzed Michael reactions.
Ye W, et al.
Tetrahedron Letters, 46(40), 6875-6878 (2005)
A convenient aminolysis of esters catalyzed by 1, 5, 7-triazabicyclo [4.4. 0] dec-5-ene (TBD) under solvent-free conditions.
Sabot C, et al.
Tetrahedron Letters, 48(22), 3863-3866 (2007)
Structural and spectroscopic studies of the 1: 1 complex of lasalocid acid with 1, 5, 7-triazabicyclo [4.4. 0] dec-5-ene.
Huczynski A, et al.
Journal of Molecular Structure, 875(1), 501-508 (2008)
P-C Bond formation via direct and three-component conjugate addition catalyzed by 1, 5, 7-triazabicyclo [4.4. 0] dec-5-ene (TBD).
Jiang Z, et al.
Tetrahedron Letters, 48(1), 51-54 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service