Skip to Content
Merck
All Photos(1)

Key Documents

69898

Sigma-Aldrich

Bromobimane

suitable for fluorescence, BioReagent, ≥95% (HPCE)

Synonym(s):

Monobromobimane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BrN2O2
CAS Number:
Molecular Weight:
271.11
Beilstein:
4430959
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥95% (HPCE)

form

solid

mp

152-154 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
acetonitrile: soluble
methanol: soluble

fluorescence

λex 390 nm; λem 478 nm in 0.1 M phosphate pH 7.5 (after derivatization with glutathione)
λex 398 nm

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CC1=C(C)C(=O)N2N1C(CBr)=C(C)C2=O

InChI

1S/C10H11BrN2O2/c1-5-7(3)12-8(4-11)6(2)10(15)13(12)9(5)14/h4H2,1-3H3

InChI key

AHEWZZJEDQVLOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromobimane, also called probe for thiols, is a fluorescent reagent activated upon a photolysis reaction. Bromobimane in solution reacts with small thiol groups (e.g., GSH) and with reactive protein thiol groups (e.g., hemoglobin). The reaction of Bromobimane with thiols is of second-order and dependent on pH. And, upon reacting with thiolate, it activates the water-soluble fluorescent product for detection.

Application

Bromobimane is used for the determination of thiols by the HPLC method. It is suitable as a pre-column derivatization agent for fluorometric determination of 2,3-dimercaptopropane-1-sulfonic acid and other dithiols. Bromobimane has been used as a fluorescent label in studying oligomycin-sensitive ATPase from beef heart mitochondria.

Features and Benefits

The benefits of using Bromobimane for the analysis of low molecular weight thiols are: Highly selective.
  • Rapid reactivity.
  • Ease of separation of derivatives by reverse HPLC.
  • Ability to penetrate cells.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Fluorescent thiol-specific reagent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Maiorino et al.
Journal of chromatography, 374(2), 297-310 (1986-01-24)
The increasing therapeutic use of dithiol metal binding agents, such as 2,3-dimercaptopropane-1-sulfonic acid (DMPS), has stimulated the need for a sensitive and selective method for their determination in biological fluids. A method has now been developed in which DMPS was
P C Chinn et al.
Analytical biochemistry, 159(1), 143-149 (1986-11-15)
A highly sensitive and specific assay for Escherichia coli thioredoxin was developed using the thiol-specific reagent monobromobimane. Treatment of dithiothreitol-reduced thioredoxin with an excess of monobromobimane in Tris buffer (pH 8.0, 23 degrees C) for 30 min resulted in the
Michael Hall et al.
Proteomics, 10(5), 987-1001 (2010-01-06)
The light-dependent regulation of stromal enzymes by thioredoxin (Trx)-catalysed disulphide/dithiol exchange is known as a classical mechanism for control of chloroplast metabolism. Recent proteome studies show that Trx targets are present not only in the stroma but in all chloroplast
Florencia Sardi et al.
Analytical biochemistry, 435(1), 74-82 (2013-01-09)
A method based on the differential reactivity of thiol and thiolate with monobromobimane (mBBr) has been developed to measure nucleophilicity and acidity of protein and low-molecular-weight thiols. Nucleophilicity of the thiolate is measured as the pH-independent second-order rate constant of
Joachim Volk et al.
Dental materials : official publication of the Academy of Dental Materials, 25(12), 1556-1563 (2009-09-01)
Camphorquinone (CQ) is cytotoxic in cell cultures. The mechanism of this toxic action, however, is not yet clearly understood. Aim of this investigation was to analyze the effects of non-irradiated CQ on intracellular formation of reactive oxygen species (ROS), intracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service