Skip to Content
Merck
All Photos(1)

Key Documents

48230

Sigma-Aldrich

D-Galacto-D-mannan from Ceratonia siliqua

~95% (HPLC)

Synonym(s):

Carob galactomannan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Ceratonia siliqua

Assay

~95% (HPLC)

form

powder

mol wt

Mr ~200000

loss

≤5% loss on drying

color

white

solubility

1 g/L at 20 °C

cation traces

Ca: ≤1000 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤100 mg/kg
K: ≤1000 mg/kg
Li: ≤5 mg/kg
Mg: ≤200 mg/kg
Mn: ≤5 mg/kg
Na: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤10 mg/kg
Ti: ≤5 mg/kg
Zn: ≤50 mg/kg

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

D-Galacto-D-mannan is a plant cell wall polysaccharide useful as a reference compound in procedures that analyse plant cell wall polysaccharide composition. D-Galacto-D-mannan is a potential anti-oxidant that protects plant cell walls.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Reserve carbohydrate from the endosperm walls of some legume seeds. Galactomannans: properties and applications; Improved resolution of DNA fragments in polysaccharide-supplemented agarose gels

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Khanna et al.
Indian journal of medical microbiology, 31(1), 34-39 (2013-03-20)
Diagnosis of invasive aspergillosis (IA) in immunocompromised patients using galactomannan ELISA (GM-ELISA) has shown variable sensitivity and specificity. To assess the diagnostic performance of GM-ELISA and analyze the effect of decreasing the cut off value, neutropenia, antifungals and piperacillin-tazobactam (PTZ).
Soo-Han Choi et al.
Pediatric blood & cancer, 60(2), 316-322 (2012-10-09)
Invasive aspergillosis (IA) is a major cause of morbidity and mortality in immunocompromised children. We investigated the usefulness of an Aspergillus galactomannan (GM) antigen assay as a diagnostic tool for IA in pediatric cancer patients and hematopoietic cell transplantation (HCT)
Nathan P Wiederhold et al.
Journal of clinical microbiology, 51(2), 459-465 (2012-11-24)
Interest in lateral-flow devices (LFDs) as potential point-of-care assays for the diagnosis of infectious diseases has increased. Our objective was to evaluate the interlaboratory and interstudy reproducibility and the effects of antifungal therapy on an LFD developed for invasive pulmonary
P Lewis White et al.
Clinical and vaccine immunology : CVI, 20(4), 636-638 (2013-02-08)
Diagnostic galactomannan (GM) enzyme immunoassay (EIA) testing is formally validated only for serum, though in practice, plasma is occasionally tested. It is assumed, but not confirmed, that results will be comparable to those for serum. GM EIA when testing plasma
Valentina Salas et al.
Antimicrobial agents and chemotherapy, 57(3), 1532-1534 (2013-01-09)
We evaluated the efficacy of voriconazole against nine strains of Aspergillus terreus with different MICs (0.12 to 4 μg/ml) by using a murine model. Markers of efficacy included survival, tissue burden, galactomannan antigenemia, and drug serum levels. Voriconazole was especially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service