Skip to Content
Merck
All Photos(3)

Key Documents

D87201

Sigma-Aldrich

2-Chloro-N,N-diethylethylamine hydrochloride

99%

Synonym(s):

2-(Diethylamino)ethyl chloride hydrochloride, 2-Chlorotriethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCH2CH2Cl · HCl
CAS Number:
Molecular Weight:
172.10
Beilstein:
3591991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

mp

208-210 °C (lit.)

SMILES string

Cl.CCN(CC)CCCl

InChI

1S/C6H14ClN.ClH/c1-3-8(4-2)6-5-7;/h3-6H2,1-2H3;1H

InChI key

RAGSWDIQBBZLLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-N,N-diethylethylamine hydrochloride can be used as an alkylating reagent for the synthesis of:
  • Substituted oxindole derivatives to be used as growth hormone secretagogs.
  • 1-Substituted-5,6-dinitrobenzimidazoles with antimicrobial and antiprotozoal activities.
  • Thiophene containing triarylmethane (TRAM) derivatives to be used as antitubercular agents.
  • Tris(2-(diethylamino)ethyl)amine to be used as a catalyst in combination with CuBr (Et6TREN/CuBr) for atom transfer radical polymerization of n-butyl acrylate.

It can also be used for the derivatization of chitosan to incorporate aminofunctionality onto it at C-6 position.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxindole derivatives as orally active potent growth hormone secretagogues.
Tokunaga T, et al.
Journal of medicinal chemistry, 44(26), 4641-4649 (2001)
Reactive oxygen species scavenging activity of aminoderivatized chitosan with different degree of deacetylation.
Je JY, et al.
Bioorganic & Medicinal Chemistry, 14(17), 5989-5994 (2006)
Thiophene containing triarylmethanes as antitubercular agents.
Parai MK, et al.
Bioorganic & Medicinal Chemistry Letters, 18(1), 289-292 (2008)
New amine-based tripodal copper catalysts for atom transfer radical polymerization.
Inoue Y and Matyjaszewski K
Macromolecules, 37(11), 4014-4021 (2004)
Ahmed M Omer et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-21)
An effective drug nanocarrier was developed on the basis of a quaternized aminated chitosan (Q-AmCs) derivative for the efficient encapsulation and slow release of the curcumin (Cur)-drug. A simple ionic gelation method was conducted to formulate Q-AmCs nanoparticles (NPs), using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service