Skip to Content
Merck
All Photos(1)

Key Documents

378R-1

Sigma-Aldrich

Cadherin-17 (SP183) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP183, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (378R-14)
vial of 0.5 mL concentrate (378R-15)
bottle of 1.0 mL predilute (378R-17)
vial of 1.0 mL concentrate (378R-16)
bottle of 7.0 mL predilute (378R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

colorectal carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... CDH17(1015)

General description

Cadherin-17, also called liver-intestinal (LI) cadherin or human peptide transporter-1, is a member of the cadherin superfamily. Unlike some classic cadherins, such as E-, N-, and P-cadherins, cadherin-17 has seven cadherin repeats instead of five within the extracellular domain and only 20 amino-acid residues in the cytoplasmic domain. The markedly short cytoplasmic domain lacks homology with other cadherins and the adhesive function of cadherin-17 is not dependent on association with other cytoplasmic proteins. The subcellular distribution of cadherin-17 is also different from classic cadherins. In intestinal epithelial cells, E-cadherin is concentrated in adherens junctions whereas cadherin-17 is evenly distributed along the lateral contact area. Human normal tissues that are strongly stained with cadherin-17 include appendicular epithelium, colonic epithelium, and small intestinal epithelium. Other normal human tissues are not stained with cadherin-17. The results above indicate cadherin-17 can be used as a marker for identification of primary sites of tumors. In-house studies have shown that cadherin-17 expression is usually diffuse and strong in colorectal adenocarcinomas, whereas it is usually focal or scattered in adenocarcinomas of the stomach, pancreas and bile duct, and is virtually absent in tumors of other anatomic sites.

Quality


IVD

IVD

IVD

RUO

Linkage

Cadherin-17 Positive Control Slides, Product No. 378S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Cheng Su et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 21(11), 1379-1386 (2008-06-17)
Cadherin-17, also called liver-intestine cadherin, is a calcium-dependent transmembrane glycoprotein that mediates cell-cell adhesion in intestinal epithelium. Expression of cadherin-17 was reported in gastric, pancreatic, and colorectal adenocarcinomas but not in other tumors. Whether cadherin-17 can be used as a
R Gessner et al.
Annals of the New York Academy of Sciences, 915, 136-143 (2001-02-24)
The cadherin superfamily comprises a large number of cell adhesion molecules, several of which are expressed in the gastrointestinal tract. LI-cadherin represents a novel type of cadherin within the cadherin superfamily distinguished from other cadherins by structural and functional features

Articles

Colorectal cancer is the third most common cancer in both men and women. An estimated 136,000 cases of colorectal cancer are expected to occur in 2016.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service