Skip to Content
Merck
All Photos(1)

Key Documents

198M-1

Sigma-Aldrich

CD123 (6H6) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

6H6, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (198M-14)
vial of 0.5 mL concentrate (198M-15)
bottle of 1.0 mL predilute (198M-17)
vial of 1.0 mL concentrate (198M-16)
bottle of 7.0 mL predilute (198M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1κ

control

blastic plasmacytoid dendritic cell neoplasm

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Blastic plasmacytoid dendritic cell neoplasm (BPDCN), previously known as CD4+/CD56+ hematodermic neoplasm or blastic NK-cell lymphoma, is a malignant neoplasm composed of immature hematopoietic precursors of plasmacytoid dendritic cells. The most frequent manifestation is a skin lesion, bone marrow involvement, and regional lymphadenopathy. Myeloid leukemia cutis (LC), myeloid sarcoma, and large aggressive B cell lymphomas should be differentiated from BPDCN. Recently, it has been reported that these entities can be distinguished by using immunohistochemistry (IHC) in paraffin-embedded tissue sections. In this study, 23 myeloid LC and 12 BPDCN cases were evaluated using a panel of antibodies against CD123, TCL1, CD4, CD56, MPO and CD33; with results as follows: anti-CD123 stained 4 cases (17%) of myeloid LC and 10 cases (83%) of BPDCN; anti-TCL-1 stained 2 cases (9%) of myeloid LC and 9 (82%) of 11 cases of BPDCN; anti-CD4 stained 2 cases (9%) of LC and all 12 cases (100%) of BPDCN; anti-CD56 stained 12 cases (52%) of LC and all 12 cases (100%) of BPDCN; anti-myeloperoxidase stained 7 cases (30%) of LC and 0 cases (0%) of BPDCN. Anti-CD33 was not helpful; it stained 18 (78%) cases of LC and 11 cases (92%) of BPDCN. The results indicated that a panel that includes antibodies against CD4, CD56, CD123, and TCL-1 can appropriately distinguish between myeloid LC and BPDCN.
CD123 IHC expression has been studied in 157 acute myeloid leukemia (AML) bone marrow biopsies and/or marrow particle preparations, and correlated with the morphologic, immunophenotypic, and cytogenetic features and with the presence of FLT3-ITD and NPM1 mutations. CD123 IHC expression has been seen in 40% of AML, across a wide spectrum of 2008 World Health Organization subtypes and was most frequent within the intermediate risk group. Compared with CD123 IHC negative AML, CD123 IHC positive AML demonstrated higher marrow blast percentages (median 69%), monocytic differentiation (33/63 cases), and CD34 negativity (29/63 cases). 83% (25/30) FLT3-ITD-mutated AML were CD123+ and 62% (18/29) NPM1-mutated cases were CD123 IHC+ (P=0.0052). CD123 IHC+AML presents with characteristic pathologic features, some of which may be related to underlying FLT3-ITD and/or NPM1 mutations.

Quality


IVD

IVD

IVD

RUO

Linkage

CD123 Positive Control Slides, Product No. 198S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danielle M P Cronin et al.
American journal of clinical pathology, 137(3), 367-376 (2012-02-18)
Myeloid leukemia cutis (LC) and blastic plasmacytoid dendritic cell neoplasm (BPDCN) are morphologically indistinguishable malignancies that frequently manifest in the skin. Separating myeloperoxidase-negative LC from BPDCN may be particularly challenging. We identified a panel of immunohistochemical stains to distinguish myeloid
Marian Rollins-Raval et al.
Applied immunohistochemistry & molecular morphology : AIMM, 21(3), 212-217 (2012-08-24)
FLT3-ITD and NPM1 mutation testing in acute myeloid leukemia (AML) plays an important role in prognostic risk stratification, especially within the intermediate cytogenetic risk group. Molecular studies require adequate fresh material and are typically performed on a dedicated aspirate specimen
Facchetti F, et al.
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 145-147 (2008)
Facchetti F, et al.
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 145-147 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service