Skip to Content
Merck
All Photos(1)

Key Documents

294R-1

Sigma-Aldrich

BOB.1 (SP92) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP92, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (294R-14)
vial of 0.5 mL concentrate (294R-15)
bottle of 1.0 mL predilute (294R-17)
vial of 1.0 mL concentrate (294R-16)
bottle of 7.0 mL predilute (294R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

B-cell lymphoma, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... POU2AF1(5450)

General description

Expression of BOB.1/OBF.1 is restricted largely to mature B-cells. Germinal center B-cells normally demonstrate strong staining for BOB.1, as do mantle-zone B-cells, and plasma cells. Analyses of BOB.1/OBF.1 expression in a variety of established B-cell lines representing different stages of B-cell development has suggested a constitutive, B-cell-specific expression pattern. Because they are germinal center derived, L&H cells in nodular lymphocyte predominant Hodgkin lymphoma are consistenty immunoreactive for BOB.1. Conversely, the Hodgkin/Reed- Sternberg cells in classical Hodgkin lymphoma either do not express both (80%) or express only one (20%) of the two proteins. In B-cell lymphomas, the highest expression levels for BOB.1/OBF.1 are reported in follicular center lymphomas, diffuse large B-cell lymphomas and Burkitt lymphomas.B-CLL, MALT-type, and mantle cell lymphomas score negative or display an heterogenous/weaker reactivity.10-11 Studies have suggested that anti-BOB. has a high predictive value in discriminating primary mediastinal B-cell lymphoma from classical Hodgkin disease.Approximately 50% of acute myeloid leukemias express BOB.1.

Linkage

BOB.1 Positive Control Slides, Product No. 294S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Greiner et al.
The American journal of pathology, 156(2), 501-507 (2000-02-10)
The BOB.1/OBF.1/OCAB.1 protein is a lymphocyte-specific transcriptional coactivator. It interacts with the Oct1 and Oct2 transcription factors and contributes to the transcriptional activity of octamer motifs. The analysis of established B cell lines had suggested that BOB.1/OBF.1 is constitutively expressed
Ana-Isabel Sáez et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 15(3), 211-220 (2002-03-21)
Oct1 and Oct2 are transcription factors of the POU homeo-domain family that bind to the Ig gene octamer sites, regulating B-cell-specific genes. The function of these transcription factors is dependent on the activity of B-cell-restricted coactivators such as BOB.1/OBF.1. Independent
Susanne Annette Steimle-Grauer et al.
Virchows Archiv : an international journal of pathology, 442(3), 284-293 (2003-03-21)
The World Health Organization (WHO) classification of Hodgkin lymphoma (HL) distinguishes two types: Classical Hodgkin lymphoma (CHL) and nodular lymphocyte predominant Hodgkin lymphoma (NLPHL). Both groups have in common that they mostly derive from B cells with rare classical cases
Stefano A Pileri et al.
The American journal of pathology, 162(1), 243-253 (2003-01-01)
Although primary mediastinal (thymic) large B-cell lymphoma has been primarily studied, its precise phenotype, molecular characteristics, and histogenesis are still a matter of debate. The International Extranodal Lymphoma Study Group collected 137 such cases for extensive pathological review. Histologically, the
Christoph Loddenkemper et al.
The Journal of pathology, 202(1), 60-69 (2003-12-25)
It has previously been demonstrated that in cultured and in situ tumour cells of classical Hodgkin lymphoma (cHL), the immunoglobulin (Ig) promoter is inactive and its transcription factors Oct2 and/or BOB.1/OBF.1 are down-regulated. In this study, the analysis of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service