Skip to Content
Merck
All Photos(1)

Documents

27487

Sigma-Aldrich

Citric acid

BioUltra, anhydrous, ≥99.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021801
PubChem Substance ID:
NACRES:
NA.25

Quality Level

product line

BioUltra

Assay

≥99.5% (T)

form

powder or crystals

expl. lim.

8 %, 65 °F

technique(s)

cell culture | hybridoma: suitable

impurities

insoluble matter, passes filter test

ign. residue

≤0.05% (as SO4)

loss

≤0.5% loss on drying, 110 °C

pH

1.0-2.0 (25 °C, 1 M in H2O)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5 mg/kg
oxalate (C2O42-): ≤0.05%
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg
tartrate (C4H4O62-): ≤0.20%

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λmax

260 nm (Amax: ≤0.20)
280 nm (Amax: ≤0.10)

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.20
λ: 280 nm Amax: 0.10

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

General description

Citric acid is a buffer component for triple-column amino acid analysis. It is an optimal buffer for dissociation of radioiodinated antibodies from viable murine tumor cells.

Application


  • Enhancing soil microbiomes: Citric acid has been applied to modify soil bacterial communities in saline mudflats, demonstrating its utility in environmental biotechnology for sustainable ecosystem management (Liu et al., 2024).

  • Biodegradable packaging in food industry: In food chemistry, citric acid is used in conjunction with polyvinyl alcohol for developing antimicrobial packaging, highlighting its potential in food safety and preservation (Xu et al., 2024).

  • Nanomaterial synthesis for engineering applications: Citric acid serves as a key component in the eco-friendly synthesis of hybrid nanomaterials for supercapacitors, underpinning its role in advancing materials science and engineering (Teixeira et al., 2024).

  • Environmental science and pollution control: Research utilizes citric acid to study the co-transport of pharmaceuticals and nanoparticles in environmental systems, aiding in the understanding of contaminant dynamics and mitigation strategies (Cui et al., 2024).

  • Food science and technology: Citric acid is employed in the formulation of vegan gummies, indicating its versatility in food product development and its impact on textural properties (Rawat et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T N Ferraro et al.
Analytical biochemistry, 143(1), 82-94 (1984-11-15)
A highly resolving triple-column amino acid analyzer with fluorometric detection is described. The reliability of this technique was evaluated and it was used in a baseline investigation of amino acids and related compounds in human cerebrospinal fluid (CSF). The procedure
R Ehrlich et al.
Journal of immunological methods, 26(4), 345-353 (1979-01-01)
Several low pH buffers were used in order to dissociate radioiodinated antibodies from sensitized living tumor cells. Three buffers were further tested for their dissociation abilities under different conditions of time and temperature, and for their influence on the eluted
Maria José Garcia-Fernandez et al.
Biofouling, 29(3), 261-271 (2013-02-27)
Prevention and management of wound infections receive a lot of attention, since the presence of micro-organisms interferes with the wound-healing process. The aim of this work was to use cyclodextrins (CDs) to endow hydrogels and gauzes with the ability to
Felix Schiele et al.
mAbs, 7(5), 871-880 (2015-06-06)
Novel oral anticoagulants are effective and safe alternatives to vitamin-K antagonists for anticoagulation therapy. However, anticoagulation therapy in general is associated with an elevated risk of bleeding. Idarucizumab is a reversal agent for the direct thrombin inhibitor, dabigatran etexilate (Pradaxa®)
Wayne B Batchelor et al.
Circulation, 106(12), 1470-1476 (2002-09-18)
The relative anti-aggregatory effects of currently prescribed platelet glycoprotein IIb/IIIa receptor antagonists during and after percutaneous coronary intervention for acute coronary syndromes have not been established. We randomized 70 acute coronary syndrome patients undergoing percutaneous coronary intervention to receive abciximab

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service