Skip to Content
Merck
All Photos(1)

Key Documents

W302600

Sigma-Aldrich

Sodium citrate dihydrate

≥99%, FG

Synonym(s):

Sodium citrate tribasic dihydrate, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
FEMA Number:
3026
Beilstein:
6104939
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
E Number:
E 331 (iii)
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1333/2008 & 178/2002

Assay

≥99%

mp

>300 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium citrate dihydrate is used as a buffer, pH controlling agent, emulsifier, sequestrant and complexing agent in food industry.

Application

Sodium citrate dihydrate is used as a buffering, sequestering, or emulsifying agent in food and beverages. It can also be used in detergent formulations due to its rapid biodegradability characteristics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food ingredients, 129-129 (2011)
Characterization of sodium tripolyphosphate and sodium citrate dehydrate residues on surfaces
Gurses MS, et al.
Talanta, 176, 8-16 (2018)
Development of better-quality assay method for the citric acid and sodium citrate in ophthalmic/oral solutions and their application to deformulation studies
Hotha KK, et al.
American Journal of Analytical Chemistry, 5, 1249-1249 (2014)
Megan E Oest et al.
Radiation research, 181(4), 439-443 (2014-04-08)
Advanced glycation end products (AGEs) are an abnormal modification of the collagenous matrix in bone, and their accumulation contributes to alteration of mechanical properties. Using a mouse model of focal external radiotherapy, we quantified the time-dependent changes in the glycation
Christophe Chapard et al.
Molecular cell, 75(2), 224-237 (2019-06-16)
Cohesin entraps sister DNAs within tripartite rings created by pairwise interactions between Smc1, Smc3, and Scc1. Because Smc1/3 ATPase heads can also interact with each other, cohesin rings have the potential to form a variety of sub-compartments. Using in vivo cysteine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service