Merck
  • Antiseptic cyclodextrin-functionalized hydrogels and gauzes for loading and delivery of benzalkonium chloride.
Antiseptic cyclodextrin-functionalized hydrogels and gauzes for loading and delivery of benzalkonium chloride.
Biofouling (2013-02-27)
Maria José Garcia-Fernandez, Gilles Brackman, Tom Coenye, Angel Concheiro, Carmen Alvarez-Lorenzo
ABSTRACT
Prevention and management of wound infections receive a lot of attention, since the presence of micro-organisms interferes with the wound-healing process. The aim of this work was to use cyclodextrins (CDs) to endow hydrogels and gauzes with the ability to take up antiseptics and sustain their delivery for several hours. Benzalkonium chloride (BzCl) can form inclusion complexes with cross-linked CDs that regulate the release through an affinity-driven mechanism. Grafting of CDs to cotton gauzes using citric acid as the linker, at 190 °C and for 15 min, led to grafting yields of about 148%, much larger than those obtained at 180 °C or with shorter reaction times. Microbiological tests revealed that the BzCl-loaded networks can inhibit the growth of Staphylococcus epidermidis and Escherichia coli both on agar plates and in liquid medium. Furthermore, the antiseptic-loaded gauzes were able to inhibit biofilm formation by Staphylococcus aureus RN1HG pMV158GFP when applied in early stages of biofilm formation and could reduce the number of living cells in preformed biofilms grown in a chronic wound biofilm model. These findings highlight the role of CDs as main components of hydrogels and gauzes for the efficient delivery of antiseptics.
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)