All Photos(3)
About This Item
Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent
form:
powder
Recommended Products
grade
reagent
vapor pressure
<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)
product line
ReagentPlus®
Assay
≥99.0%
form
powder
pH
5-6 (25 °C, 119 g/L)
mp
734 °C (lit.)
SMILES string
[K+].[Br-]
InChI
1S/BrH.K/h1H;/q;+1/p-1
InChI key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
May be used for removal of peripheral membrane proteins.
Other Notes
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jun Pan et al.
Organic letters, 13(18), 4974-4976 (2011-08-26)
A facile Pd-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) is described. This method allows convenient access to a variety of aryl, heteroaryl, and vinyl halides in good to excellent yields and with
Kazue Kurihara
Advances in colloid and interface science, 158(1-2), 130-138 (2010-05-11)
Brush layers of polyelectrolytes, ionized chains of poly(glutamic acid) (PLGA) and poly(lysine) (PLL), prepared by the Langmuir-Blodgett method, were characterized using the surface forces measurements at various pHs, salt concentrations and chain densities. This paper reviews the major results: (1)
Pascal Miéville et al.
Magnetic resonance in chemistry : MRC, 49(11), 689-692 (2011-10-18)
As previously demonstrated by Thurber and Tycko, the peak position of (79)Br in potassium bromide (KBr) allows one to determine the temperature of a spinning sample. We propose to adapt the original design by using a compact KBr tablet placed
Keiichiro Hayashi et al.
Epilepsia, 52(5), 1010-1017 (2011-04-13)
Mutations in the SCN1A gene, which encodes the α1 subunit of voltage-gated sodium channels, cause generalized epilepsy with febrile seizures plus (GEFS+) and severe myoclonic epilepsy of infancy (SMEI). N1417H-Scn1a mutant rats are considered to be an animal model of
Hitesh Kulhari et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1511-1520 (2015-05-03)
Docetaxel (DTX) is an anticancer drug that is used alone and in combination with other drugs to treat tumours. However, it suffers from the drawback of non-specific cytotoxicity. To improve the therapeutic potential of DTX, we report the synthesis of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service