Skip to Content
Merck
All Photos(1)

Key Documents

52738-U

Supelco

Supelclean PSA SPE Bulk Packing

pkg of 100 g

Synonym(s):

PSA SPE material (primary secondary amine)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

Agency

suitable for GB 5009.253-2016

Quality Level

product line

Supelclean

feature

endcapped

packaging

pkg of 100 g

extent of labeling

8.4% C
3.3% N loading

technique(s)

solid phase extraction (SPE): suitable

surface area

500 m2/g

surface coverage

surface coverage 2.35 μmol/m2

matrix

silica gel base material (irregularly shaped)

matrix active group

ethylenediamine-N-propy bonding

particle size

50 μm

pore size

70 Å pore size

pKa 

(1) 10.1, (2) 10.9

capacity

0.98-1.05 meq/g capacity

application(s)

food and beverages

separation technique

ion exchange
normal phase

General description

Retention Mechanism: Normal-phase and anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • Polymerically bonded, ethylenediamine-N-propyl phase that contains both primary and secondary amines
  • A weak anion exchanger with a pKa of 10.1 and 10.9
  • Similar to aminopropyl SPE phases (NH2) in terms of selectivity, but has a much higher capacity due to presence of secondary amine (0.98-1.05 meq/g)
  • Strong affinity and high capacity for removing fatty acids, organic acids, and some polar pigments and sugars when conducting multi-residue pesticide analysis in foods
  • Has been shown to significantly reduce matrix-enhancement effects encountered during the GC analysis of food products
  • Tested for superior cleanliness using GC-FID and GC-MS
  • Bidentate nature of ligand allows for chelation

Application

Supelclean PSA SPE Bulk Packing may be used as a sorbent in the solid-phase dispersive extraction followed by single flow injection multi-commuted system/solid-surface fluorescence spectroscopic analysis of diphenylamine in apples and pears.

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronica Padula et al.
Marine pollution bulletin, 158, 111435-111435 (2020-08-06)
Phthalates, plastic-derived contaminants, are of increasing global concern. This study quantified phthalates in seabirds collected across >1700 km of the Aleutian Islands, Alaska, and contributes to a body of knowledge on plastic contaminants in marine wildlife. We measured six phthalate congeners
Rapid determination of diphenylamine residues in apples and pears with a single multicommuted fluorometric optosensor
Garcia-Reyes FJ, et al.
Journal of Agricultural and Food Chemistry, 53(26), 9874-9878 (2005)

Articles

SPE retention mechanism in this case is based on the electrostatic attraction of charged functional groups of the analyte(s) to oppositely charged functional groups on the sorbent.

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Related Content

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service