Skip to Content
Merck
All Photos(3)

Key Documents

P0838

Sigma-Aldrich

Potassium bromide

BioXtra, ≥99.0%

Synonym(s):

Bromide salt of potassium, KBr

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050405
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0%
form:
powder
solubility:
H2O: 1 M, clear, colorless
Pricing and availability is not currently available.

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

Quality Level

product line

BioXtra

Assay

≥99.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

mp

734 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
Ba: ≤0.002%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.15%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for removal of peripheral membrane proteins.
Potassium bromide has been used in the sample preparation for fourier transform infrared spectroscopy (FT-IR).[1][2][3] It has also been used as a supplement in Wilbrink Selective Davis (WSD) medium.[4]

Biochem/physiol Actions

Potassium bromide (KBr) is used as an anticonvulsant and sedative. KBr is used for optical windows and prisms. KBr is transparent in the wide wavelength range from near ultraviolet to long wave infrared. It is employed in the sample preparation for infrared transmission spectra.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of luminescent graphene quantum dots on trypsin activity
Tabish TA, et al.
International journal of nanomedicine, 13, 1525-1525 (2018)
Bioassay to Study the Attachment of Xanthomonas albilineans on Sugarcane Leaves
Mensi I, et al.
Bio-protocol, 7(2) (2017)
Isolation and characterization of phthalates from Brevibacterium mcbrellneri that cause cytotoxicity and cell cycle arrest
Rajamanikyam M, et al.
EXCLI Journal , 16, 375-375 (2017)
Health & Drugs: Disease, Prescription & Medication (2014)
Eilish Broderick et al.
Journal of colloid and interface science, 298(1), 154-161 (2006-01-18)
The characterisation of a novel amphiphilic material, Alg-C4, produced from butanol linked by esterification to alginate is presented. The novel derivative retains the gelling and non-toxic properties of native alginate. FTIR spectra of Alg-C4 contained the characteristic hydroxyl and carboxyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service