Skip to Content
Merck
All Photos(9)

Key Documents

746371

Sigma-Aldrich

Sodium iodide

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
grade:
ACS reagent
anhydrous
form:
powder

grade

ACS reagent
anhydrous

Quality Level

vapor density

>1 (vs air)

product line

Redi-Dri

Assay

≥99.5%

form

powder

quality

free-flowing

impurities

≤0.002% N compounds
≤0.01% insolubles

pH

6.0-9.0 (25 °C, 5%)

mp

661 °C (lit.)

anion traces

Cl- and Br-: ≤0.01%
iodate (IO3-): ≤3 ppm
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤5 ppm
K: ≤0.01%
Mg: ≤0.001%
heavy metals (as Pb): ≤5 ppm

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium iodide is an inorganic sodium salt that is commonly used as a source of iodide ions in organic synthesis. It is used in the conversion of alkyl halides and sulfonates into alkyl iodides and aryl iodides via nucleophilic displacement reactions. Additionally, NaI is also used in combination with oxidizing agents for the iodination of organoboranes, phenols, and aromatic compounds.

Application

Sodium iodide can be used in the following:      
  • NaI-AlCl3 is used as a dehydrating agent in the Beckmann rearrangement of ketoximes to anilides.      
  • NaI-cerium chloride supported on silica gel is used as a catalyst for the synthesis of 2,3-dihydro-1H-1,5-benzodiazepines by the reaction of o-phenylenediamines and ketones.      
  • NaI-dichlorodimethylsilane catalytic system is used for the reduction of diarylmethanols to corresponding diarylmethanes.

Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders     
  • Safety – Eliminates tools of force used to break up clumps
  • Ecology – Reduces waste as it does not stick to the packaging      
  • Economy – Faster preparation and solvation increase productivity and reduce costs      
  • Assured Quality – Excellent, expert-tested quality with no anti-caking agents      
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuro Isoda et al.
Clinical nuclear medicine, 39(8), 680-684 (2014-07-01)
Many cases of unexpected radioiodine uptake have been reported, including physiological uptake in healthy tissue and in both benign and malignant nonthyroidal lesions. However, iodine uptake in the uterus has not been well assessed. In this article, we systemically analyzed
Anke de Vries et al.
Investigative radiology, 50(4), 297-304 (2015-01-01)
The objective of this study was to investigate the feasibility and the accuracy of spectral computed tomography (spectral CT) to determine the tissue concentrations and localization of high-attenuation, iodine-based contrast agents in mice. Iodine tissue concentrations determined with spectral CT
Dafu Yu et al.
Hormones and behavior, 69, 106-115 (2015-01-28)
Thyroid hormone disorders have long been linked to depression, but the causal relationship between them remains controversial. To address this question, we established rat models of hypothyroidism using (131)iodine ((131)I) and hyperthyroidism using levothyroxine (LT4). Serum free thyroxine (FT4) and
Kerstin Knoop et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 56(4), 600-606 (2015-03-07)
The tumor-homing property of mesenchymal stem cells (MSCs) allows targeted delivery of therapeutic genes into the tumor microenvironment. The application of sodium iodide symporter (NIS) as a theranostic gene allows noninvasive imaging of MSC biodistribution and transgene expression before therapeutic
Yayoi S Kikkawa et al.
Journal of translational medicine, 12, 280-280 (2014-10-05)
A cochlear implant (CI) is an artificial hearing device that can replace a damaged cochlea. The present study examined the use of growth factor-eluting gelatin hydrogel coatings on the electrodes to minimize inner ear trauma during electrode insertion. Insulin-like growth

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service