Skip to Content
Merck
All Photos(4)

Key Documents

383112

Sigma-Aldrich

Sodium iodide

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
chunks or granules
crystalline powder
powder or crystals

grade

ACS reagent

vapor density

>1 (vs air)

vapor pressure

~1 hPa ( 767 °C)

Assay

≥99.5%

form

chunks or granules
crystalline powder
powder or crystals

impurities

≤0.002% N compounds
≤0.01% insolubles

pH

6.0-9.0 (25 °C, 5%)

mp

661 °C (lit.)

anion traces

Cl- and Br-: ≤0.01%
iodate (IO3-): ≤3 ppm
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤5 ppm
K: ≤0.01%
Mg: ≤0.001%
heavy metals (as Pb): ≤5 ppm

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium iodide is a water soluble iodide salt.

Application

Sodium iodide was used as a component in the preparation of Mayer′s hematoxylin stain solution.
It may be used in the following processes:
  • Precursor in the polymerization of butyl acrylate.
  • Chaotropic agent in the extraction of DNA.
  • Deprotecting agent in the removal of N-tert-butyloxycarbonyl group in amino acids.
  • Water-soluble fluorescence quenching reagent.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium iodide as a novel, neutral and facile deprotecting reagent of N-tert-butyloxycarbonyl groups in peptide chemistry.
Ham J, et al. et al.
Protein and peptide letters, 5(5), 257-258 (1998)
Isolation and characterization of mesenchymal stem cells.
Odabas SA, et al.
Bone Marrow and Stem Cell Transplantation., 1109, 47-63 (2014)
Use of sodium iodide as the precursor to the control agent in ab initio emulsion polymerization.
Tonnar J and Lacroix-Desmazes P.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 120(7), 1314-1317 (2008)
Oxidative stress in the aging rat heart is reversed by dietary supplementation with (R)-a-lipoic acid.
Suh JH, et al.
Faseb Journal, 15(3), 700-706 (2001)
Poly(propyleneimine) Dendrimers as pH-Sensitive Controlled-Release Systems.
Pistolis G, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 5(5), 1440-1444 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service