Skip to Content
Merck
All Photos(2)

Key Documents

21004

Supelco

Cesium iodide

analytical standard, suitable for mass spectrometry (MS)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsI
CAS Number:
Molecular Weight:
259.81
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

analytical standard

Assay

99.999%

technique(s)

mass spectrometry (MS): suitable

mp

626 °C (lit.)

density

4.51 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[I-].[Cs+]

InChI

1S/Cs.HI/h;1H/q+1;/p-1

InChI key

XQPRBTXUXXVTKB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cesium iodide is a simple ionic salt. It has the capacity to undergo second order transformation from cubic B2 structure to body-centered tetragonal structure at low pressures. It is mostly used as input screens of X-ray image intensifiers.

Application

Cesium iodide has been used as calibration standard during native electrospray ionization time-of-flight mass spectrometry (ESI-TOF MS) analysis for measuring protein samples, from protein aggregates.
Suitable as standard for electrospray ionisation MS (ESI)[1][2]

Features and Benefits

Frequently used in devices such as phosphor screens for medical imaging, scintillators, calorimeters and a variety of particle detectors.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.F. Anacleto et al.
Org. Mass Spectrom., 27, 660-660 (1992)
Wei Zhao et al.
Medical physics, 31(9), 2594-2605 (2004-10-19)
Columnar structured cesium iodide (CsI) scintillators doped with Thallium (Tl) have been used extensively for indirect x-ray imaging detectors. The purpose of this paper is to develop a methodology for systematic investigation of the inherent imaging performance of CsI as
P Thibault et al.
Rapid communications in mass spectrometry : RCM, 5(10), 484-490 (1991-10-01)
The direct coupling of capillary electrophoresis (CE) and mass spectrometry, combined with ionspray ionization using a coaxial capillary arrangement, is described. The CE/mass spectrometer interface is shown to be effective for the analysis of native and tryptic peptides and of
Başak Kükrer et al.
Pharmaceutical research, 27(10), 2197-2204 (2010-08-04)
The aim of this study was to develop a method to characterize intact soluble monoclonal IgG1 antibody (IgG) oligomers by mass spectrometry. IgG aggregates (dimers, trimers, tetramers and high-molecular-weight oligomers) were created by subjecting an IgG formulation to several pH
E Knittle et al.
Science (New York, N.Y.), 223(4631), 53-56 (1984-01-06)
Cesium iodide, a simple ionic salt at low pressures, undergoes a second-order transformation at 40 gigapascals (400 kilobars) from the cubic B2 (cesium chloride-type) structure to the body-centered tetragonal structure. Also, the energy gap between valence and conduction bands decreases

Articles

MALDI-TOF MS is useful for monitoring complete conjugation.  Functionalized polymers are useful for creating polymer conjugates for diverse applications.

MALDI-TOF MS is useful for monitoring complete conjugation.  Functionalized polymers are useful for creating polymer conjugates for diverse applications.

MALDI-TOF MS is useful for monitoring complete conjugation.  Functionalized polymers are useful for creating polymer conjugates for diverse applications.

MALDI-TOF MS is useful for monitoring complete conjugation.  Functionalized polymers are useful for creating polymer conjugates for diverse applications.

See All

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service