Skip to Content
Merck
All Photos(1)

Key Documents

49770

Sigma-Aldrich

Glycerol

puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

viscous liquid

autoignition temp.

698 °F

quality

dist.

expl. lim.

2.6-11.3 %

technique(s)

electrophoresis: suitable

impurities

acrolein and glucose, in accordance
substances darkened by H2SO4, in accordance
≤0.05% fatty acid esters (as butyric acid)
≤0.1% water

ign. residue

≤0.005%

refractive index

n20/D 1.474 (lit.)

pH

5.5-8

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
NH4+: ≤5 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol is widely used in industries based on food, polymer, resins, tobacco, detergents, explosives, drugs and personal care products. It is most commonly employed cheap feedstock in the chemical industry. Its transformation to various commercial value-added chemicals has been proposed. A fast enzymatic assay for the determination of glycerol based on its reaction with ATP has been described.

Application

Glycerol has been used in the synthesis of poly(glycerol sebacate) (PGS) pre-polymer, which is required for the fabrication of PGS scaffolds. It may be employed to protect the human red blood-cells from damage during the process of freezing and thawing.

greener alternative product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid enzymatic assay for glycerol.
Garland PB and Randle PJ.
Nature, 196, 987 - 988 (1962)
The mechanism of the protective action of glycerol against haemolysis by freezing and thawing.
J E LOVELOCK
Biochimica et biophysica acta, 11(1), 28-36 (1953-05-01)
Nafiseh Masoumi et al.
Journal of biomedical materials research. Part A, 101(1), 104-114 (2012-07-25)
Microfabricated poly(glycerol sebacate) (PGS) scaffolds may be applicable to tissue engineering heart valve leaflets by virtue of their controllable microstructure, stiffness, and elasticity. In this study, PGS scaffolds were computationally designed and microfabricated by laser ablation to match the anisotropy
Mario Pagliaro et al.
Angewandte Chemie (International ed. in English), 46(24), 4434-4440 (2007-05-02)
Today, industrial plants that produce glycerol are closing down and others are opening that use glycerol as a raw material, owing to the large surplus of glycerol formed as a by-product during the production of biodiesel. Research efforts to find
Tian Qiu et al.
Nature communications, 5, 5119-5119 (2014-11-05)
Biological microorganisms swim with flagella and cilia that execute nonreciprocal motions for low Reynolds number (Re) propulsion in viscous fluids. This symmetry requirement is a consequence of Purcell's scallop theorem, which complicates the actuation scheme needed by microswimmers. However, most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service