Skip to Content
Merck
All Photos(1)

Key Documents

G9012

Sigma-Aldrich

Glycerol

≥99.5%

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
50161901
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.5%

form

viscous liquid

autoignition temp.

698 °F

technique(s)

immunofluorescence: suitable

impurities

≤0.20% Water (Karl Fischer)

color

colorless

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

solubility

water: 1 mL/mL, clear, colorless to faintly yellow

density

1.25 g/mL (lit.)

cation traces

heavy metals (as Pb): ≤5 ppm

λ

1 cm path

UV absorption

λ: 205 nm Amax: 1.0
λ: 225 nm Amax: 0.40
λ: 280 nm Amax: 0.10
λ: 320 nm Amax: 0.04
λ: 340 nm Amax: 0.01
λ: 400 nm Amax: 0.01

suitability

suitable for component for culture media

application(s)

lipidomics
liquid formulation
sample preservation

storage temp.

room temp

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol is odourless, colorless, viscous in nature, and exists as a sweet tasting liquid. It can be derived naturally as well as from petrochemical feedstock. Glycerol has a wide variety of applications, and is one of the most valuable and versatile chemical substances in nature. It can be used as an emollient, solvent, sweetening agent, in pharmaceutical formulations, cosmetics, foodstuffs and toiletries. It is very stable and can be easily stored under normal temperature; also it is non-irritating and has no adverse impact to the environment.

Application

Glycerol has been used as a component of mounting medium for immunofluorescence. It has also been used as additives in poly(3,4-ethylenedioxythiophene) polystyrene sulfonate (PEDOT:PSS) based inks.

Biochem/physiol Actions

Glycerol is hygroscopic in nature and is soluble in water owing to its three hydrophilic alcoholic hydroxyl groups. It can form both inter- and intramolecular hydrogen bonds, making it a very flexible molecule. The physiologic effect of glycerine is due to cell-mediated immunity, increased IgG production and increased histamine release.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Towards inkjet printable conducting polymer artificial muscles
Simaite A, et al.
Sensors and Actuators B, Chemical, 229 (2016)
Mohamad-Ali Fawal et al.
Cancer letters, 503, 129-137 (2021-02-06)
Brain tumors are a heterogeneous group of benign and malignant tumors arising from the brain parenchyma and its surrounding structures, with in general a poor clinical outcome due to high recurrence. One of the underlying causes for this somber prognostic
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
T W Chew et al.
Oncogene, 33(21), 2717-2727 (2013-07-03)
RAS and Rho small GTPases are key molecular switches that control cell dynamics, cell growth and tissue development through their distinct signaling pathways. Although much has been learnt about their individual functions in both cell and animal models, the physiological
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype

Protocols

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service