Skip to Content
Merck
All Photos(3)

Key Documents

376558

Sigma-Aldrich

Iodine

flakes, ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
EC Number:
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
184 °C (lit.)
vapor pressure:
0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

grade

reagent

vapor density

9 (vs air)

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

product line

ReagentPlus®

Assay

≥99%

form

flakes

resistivity

1.3E15 μΩ-cm

dilution

(for general lab use)

impurities

≤0.05% nonvolatile matter

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodine is a micro nutrient essential for thyroid synthesis. It is a component of Gram′s iodine which has been reported to be used in identifying cellulase-producing microorganisms. Photogeneration of iodine from anatase TiO2 has been studied.

Application

Iodine may be used as a promoter during the synthesis of conjugated molecules with benzofulvene core. It may be employed as a TLC stain for detection of perillyl alcohol.
It may be used as a catalyst in the synthesis of the following:
  • Quinoxalines by reacting 1,2-diamines with 1,2-dicarbonyl compounds under microwave condition.
  • N-Substituted pyrroles via condensation of 2,5-dimethoxy tetrahydrofuran with amines under microwave condition.
  • Disulfidation of alkenes.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine-catalyzed disulfidation of alkenes.
Wang XR and Chen F.
Tetrahedron, 67(25), 4547-4551 (2011)
Debasish Bandyopadhyay et al.
Molecules (Basel, Switzerland), 15(4), 2520-2525 (2010-04-30)
An expeditious synthesis of N-substituted pyrroles has been developed by reacting 2,5-dimethoxy tetrahydrofuran and several amines using a microwave-induced molecular iodine-catalyzed reaction under solventless conditions.
Chockalingam Karunakaran et al.
Chemistry Central journal, 5(1), 31-31 (2011-06-18)
The crystal structures of different forms of TiO2 and those of BaTiO3, ZnO, SnO2, WO3, CuO, Fe2O3, Fe3O4, ZrO2 and Al2O3 nanoparticles have been deduced by powder X-ray diffraction. Their optical edges have been obtained by UV-visible diffuse reflectance spectra.
Processing cherries (Prunus avium) using supercritical fluid technology. Part 1: Recovery of extract fractions rich in bioactive compounds.
Serra AT, et al.
Journal of Supercritical Fluids, 55(1), 184-191 (2010)
Debasish Bandyopadhyay et al.
Molecules (Basel, Switzerland), 15(6), 4207-4212 (2010-07-27)
A microwave-induced iodine-catalyzed simple, rapid and convenient synthesis of different types of quinoxalines via condensation of 1,2-diamines with 1,2-dicarbonyl compounds has been accomplished with an excellent yield.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service