Skip to Content
Merck
All Photos(4)

Documents

207969

Sigma-Aldrich

Potassium iodide

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1 mmHg ( 745 °C)

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder or crystals

pH

~6.9 (20 °C, 50 g/L)

mp

681 °C (lit.)

solubility

water: soluble(lit.)

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium iodide (KI) is an inorganic potassium salt. KI is a colorless solid having cubic crystal system. It has been prepared by reacting hydroiodic acid with potassium hydroxide or potassium carbonate. Alumina loaded with potassium iodide has been employed as catalyst for the preparation of biodiesel by the transesterification reaction of soybean oil with methanol.

Application

Potassium iodide (KI) may be used in the preparation of standard iodide stock solution for the size exclusion chromatographic method for the estimation of iodide. It may be used in the spectrophotometric determination of advanced oxidation protein products (AOPP) in plasma.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 889-889 (1994)
V Witko-Sarsat et al.
Journal of immunology (Baltimore, Md. : 1950), 161(5), 2524-2532 (1998-09-02)
We previously demonstrated the presence of advanced oxidation protein products (AOPP), a novel marker of oxidative stress in the plasma of uremic patients receiving maintenance dialysis. The present study in a cohort of 162 uremic patients showed that plasma concentrations
Alumina-supported potassium iodide as a heterogeneous catalyst for biodiesel production from soybean oil.
Xie W and Li H.
J. Mol. Catal. A: Chem., 255(1), 1-9 (2006)
H B Li et al.
Journal of chromatography. A, 918(2), 335-339 (2001-06-16)
A novel method for the determination of iodide by size exclusion chromatography was established. The method was simple and highly sensitive with good precision. Iodide was converted to iodine, then sequestered with starch, and separated from the matrix using a
S C Clement et al.
Pediatric blood & cancer, 60(11), 1833-1838 (2013-07-09)
Thyroid dysfunction has been reported in up to 52% of patients 1.4 years after treatment with (131) I-Metaiodobenzylguanidine (MIBG) in children with neuroblastoma (NBL), despite the use of potassium-iodide (KI). Our aim was to investigate if the incidence and severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service