Skip to Content
Merck
All Photos(1)

Key Documents

04115

Sigma-Aldrich

Phosphorous acid solution

≥50%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PHO3
CAS Number:
Molecular Weight:
82.00
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

concentration

≥50%

impurities

≤0.001% heavy metals (as Pb)

density

1.651 g/cm3

anion traces

chloride (Cl-): ≤10 mg/kg

cation traces

Fe: ≤10 mg/kg

SMILES string

O[PH](O)=O
O[PH](O)=O

InChI

1S/H3O3P/c1-4(2)3/h4H,(H2,1,2,3)

InChI key

ABLZXFCXXLZCGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Walter Zenobi et al.
Journal of the mechanical behavior of biomedical materials, 77, 199-204 (2017-09-25)
To evaluate the bonding ability and nanoleakage of a universal adhesive applied to dentin pre-treated using a zoledronate-containing primer (zol-primer) before and after mechanical load cycling. Flat dentin surfaces obtained from human molars were assigned to one of the following
Andrew M Turner et al.
Nature communications, 9(1), 3851-3851 (2018-09-23)
Phosphorus signifies an essential element in molecular biology, yet given the limited solubility of phosphates on early Earth, alternative sources like meteoritic phosphides have been proposed to incorporate phosphorus into biomolecules under prebiotic terrestrial conditions. Here, we report on a
Qiankun Chen et al.
International journal of nanomedicine, 13, 6009-6018 (2018-10-17)
The aim of this study was to prepare transferrin (Tf) and folic acid (FA) co-modified bufalin (BF) liposomes for lung cancer treatment. In this study, (FA+Tf) BF-LPs were prepared using the high-pressure homogenization method. The EE% and DL% of prepared
Anodic oxidation mechanism of hypophosphorous and phosphorous acid on palladium.
Trasatti S and Alberti A.
Journal of Electroanalytical Chemistry, 12(3), 236-249 (1966)
Ai Nishitani et al.
Experimental animals, 69(4), 388-394 (2020-06-09)
We recently demonstrated that aspartoacylase (Aspa) and hyperpolarization-activated cyclic nucleotide-gated potassium channel 1 (Hcn1) genes were causative of essential tremor (ET) in rats. This finding was obtained using Aspaem34Kyo/Hcn1A354V double-mutant rats, but they were bred on a heterogeneous genetic background

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service