Skip to Content
Merck
All Photos(3)

Key Documents

900504

Sigma-Aldrich

Sodium carbonate

anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent (primary standard)
anhydrous

Quality Level

Agency

suitable for EPA 300

product line

Redi-Dri

Assay

99.95-100.05% dry basis

form

powder

quality

free-flowing

impurities

≤0.005% silica
≤0.01% insolubles

loss

≤1.0% loss on heating

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.02%
Fe: ≤5 ppm
K: ≤0.005%
Mg: ≤0.004%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Wen Chen et al.
American journal of physiology. Endocrinology and metabolism, 284(6), E1149-E1155 (2003-02-27)
At present, the circulating bioactivity of insulin-like growth factor I (IGF-I) is estimated by immunological measurements of IGF-I levels. However, immunoassays ignore the modifying effects of the IGF-binding proteins (IGFBPs) on the interaction between IGF-I and the IGF-I receptor (IGF-IR).
Angeles M Alonso et al.
Journal of agricultural and food chemistry, 50(21), 5832-5836 (2002-10-03)
It has been demonstrated that wine and other products derived from the grape have a high antioxidant capability; as a possible consequence of this, they may have potential benefits for health. The byproducts of the winemaking process represent a source
Mark Wahrenbrock et al.
The Journal of clinical investigation, 112(6), 853-862 (2003-09-17)
Trousseau described spontaneous, recurrent superficial migratory thrombophlebitis associated with occult cancers, and this was later correlated with disseminated microangiopathy (platelet-rich clots in small blood vessels). Trousseau syndrome often occurs with mucinous adenocarcinomas, which secrete abnormally glycosylated mucins and mucin fragments
Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)
Sandra Nilsson et al.
PloS one, 7(11), e46980-e46980 (2012-11-28)
During the intraerythrocytic development of Plasmodium falciparum, the malaria parasite remodels the host cell cytosol by inducing membranous structures termed Maurer's clefts and inserting parasite proteins into the red blood cell cytoskeleton and plasma membrane. Pf332 is the largest known

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service