Skip to Content
Merck
All Photos(1)

Documents

Y0000793

Vinyl acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acetoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH=CH2
CAS Number:
Molecular Weight:
86.09
Beilstein:
1209327
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

3 (vs air)

vapor pressure

88 mmHg ( 20 °C)

API family

vinyl acetate

autoignition temp.

801 °F

expl. lim.

13.4 %

manufacturer/tradename

EDQM

refractive index

n20/D 1.395 (lit.)

bp

72-73 °C (lit.)

mp

−93 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)OC=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vinyl acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Piperno et al.
The Journal of organic chemistry, 75(9), 2798-2805 (2010-04-17)
A new template of C-4'-truncated phosphonated nucleosides (TPCOANs) has been obtained in good yields according to two different routes which exploit the reactivity of a phosphonated nitrone. The one-step procedure based on the 1,3-dipolar cycloaddition of a phosphonated nitrone with
B Picquet-Varrault et al.
Environmental science & technology, 44(12), 4615-4621 (2010-05-25)
Vinyl acetate is widely used in industry. It has been classified as a high-production volume (HPV) chemical in the United States. To evaluate its impact on the environment and air quality, its atmospheric reactivity toward the three main tropospheric oxidants
María B Blanco et al.
Environmental science & technology, 46(16), 8817-8825 (2012-07-18)
The products formed from the reactions of OH radicals with vinyl acetate and allyl acetate have been studied in a 1080 L quartz-glass chamber in the presence and absence of NO(x) using in situ FTIR spectroscopy to monitor the reactant
Hercules V Ferreira et al.
Molecules (Basel, Switzerland), 17(8), 8955-8967 (2012-07-28)
The lipase B from Candida antarctica (Novozym 435®, CALB) efficiently catalyzed the kinetic resolution of some aliphatic secondary alcohols: (±)-4-methylpentan-2-ol, (±)-5-methylhexan-2-ol, (±)-octan-2-ol, (±)-heptan-3-ol and (±)-oct-1-en-3-ol. The lipase showed excellent enantioselectivities in the transesterifications of racemic aliphatic secondary alcohols producing the
Francesca Reineri et al.
Nature communications, 6, 5858-5858 (2015-01-06)
The advent of nuclear spins hyperpolarization techniques represents a breakthrough in the field of medical diagnoses by magnetic resonance imaging. Dynamic nuclear polarization (DNP) is the most widely used method, and hyperpolarized metabolites such as [1-(13)C]-pyruvate are shown to report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service