Skip to Content
Merck
All Photos(1)

Key Documents

83300

Sigma-Aldrich

2-Pyrrolidinone

purum, ≥98.0% (GC)

Synonym(s):

2-Pyrrolidone, Butyrolactam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO
CAS Number:
Molecular Weight:
85.10
Beilstein:
105241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.9 (vs air)

Quality Level

grade

purum

Assay

≥98.0% (GC)

form

solid

refractive index

n20/D 1.487 (lit.)

bp

245 °C (lit.)

mp

23-25 °C (lit.)

solubility

H2O: miscible (completely)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCN1

InChI

1S/C4H7NO/c6-4-2-1-3-5-4/h1-3H2,(H,5,6)

InChI key

HNJBEVLQSNELDL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

  • 2-Pyrrolidinone is used in the synthesis of N-vinylpyrrolidone, from which polyvinylpyrrolidone (PVP) is obtained.[1]
  • It can be used as a ligand in the preparation of one-dimensional coordination polymers[2] and mixed transition metal-lanthanide complexes.[3]
  • 1,2-amino alcohols can be reacted with carbon dioxide in the presence of 2-pyrrolidinone electrogenerated base to synthesize various oxazolidin-2-ones (Evans-type auxiliaries).[4]
  • It is a key starting material in the synthesis of (±)-tetraponerine T4.[5]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses, structures and magnetic properties of 1-D coordination polymers containing both dicyanamide and 2-pyrrolidone.
Sun B W, et al.
Inorganic Chemistry Communications, 4(2), 72-75 (2001)
Mixed Transition Metal? Lanthanide Complexes at High Oxidation States: Heteronuclear CeIVMnIV Clusters.
Tasiopoulos A J, et al.
Inorganic Chemistry, 46(23), 9678-9691 (2007)
The reaction of 1, 2-amino alcohols with carbon dioxide in the presence of 2-pyrrolidone electrogenerated base. New synthesis of chiral oxazolidin-2-ones.
Casadei M A, et al.
The Journal of Organic Chemistry, 65(15), 4759-4761 (2000)
2?Pyrrolidone.
Harreus A L
Ullmann's Encyclopedia of Industrial Chemistry (1993)
Electrophilic Activation of Lactams with Tf2O and Pyridine: Expedient Synthesis of (?)-Tetraponerine T4.
Charette A B, et al.
Organic Letters, 7(24), 5401-5404 (2005)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service