Merck
  • Atmospheric reactivity of vinyl acetate: kinetic and mechanistic study of its gas-phase oxidation by OH, O3, and NO3.
Atmospheric reactivity of vinyl acetate: kinetic and mechanistic study of its gas-phase oxidation by OH, O3, and NO3.
Environmental science & technology (2010-05-25)
B Picquet-Varrault, M Scarfogliero, J-F Doussin
ABSTRACT
Vinyl acetate is widely used in industry. It has been classified as a high-production volume (HPV) chemical in the United States. To evaluate its impact on the environment and air quality, its atmospheric reactivity toward the three main tropospheric oxidants (OH, NO(3), and O(3)) has been investigated. Kinetic and mechanistic experiments have been conducted at room temperature and atmospheric pressure using an indoor Pyrex simulation chamber coupled to Fourier transform infrared (FTIR) and UV-visible spectrometers. Rate constants for the reactions of vinyl acetate with OH, NO(3), and O(3) were equal to (2.3 +/- 0.3) x 10(-11), (7.3 +/- 1.8) x 10(-15), and (3.0 +/- 0.4) x 10(-18) cm(3) molecule(-1) s(-1), respectively. From these data, tropospheric lifetimes of vinyl acetate have been estimated as follows: tau(OH) = 6 h, tau(NO(3)) = 6 days, and tau(O(3)) = 5 days. This demonstrates that reaction with OH radicals is the main tropospheric loss process of this compound. From the mechanistic experiments, main oxidation products have been identified and quantified and oxidation schemes have been proposed for each studied reaction.
Product Description
Supelco
Vinyl acetate solution, certified reference material, 5000 μg/mL in acetonitrile
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Supelco
Vinyl acetate, analytical standard
Sigma-Aldrich
Vinyl acetate, CP
Vinyl acetate, European Pharmacopoeia (EP) Reference Standard