Skip to Content
Merck
All Photos(1)

Key Documents

Y0000504

Atracurium for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Atracurium besylate, 2,2′-[1,5-Pentanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-[3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinolinium]dibenzenesulfonate, BW-33A, Tracrium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H82N2O18S2
CAS Number:
Molecular Weight:
1243.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atracurium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[O-]S(=O)(=O)c1ccccc1.[O-]S(=O)(=O)c2ccccc2.COc3ccc(CC4c5cc(OC)c(OC)cc5CC[N+]4(C)CCC(=O)OCCCCCOC(=O)CC[N+]6(C)CCc7cc(OC)c(OC)cc7C6Cc8ccc(OC)c(OC)c8)cc3OC

InChI

1S/C53H72N2O12.2C6H6O3S/c1-54(22-18-38-32-48(62-7)50(64-9)34-40(38)42(54)28-36-14-16-44(58-3)46(30-36)60-5)24-20-52(56)66-26-12-11-13-27-67-53(57)21-25-55(2)23-19-39-33-49(63-8)51(65-10)35-41(39)43(55)29-37-15-17-45(59-4)47(31-37)61-6;2*7-10(8,9)6-4-2-1-3-5-6/h14-17,30-35,42-43H,11-13,18-29H2,1-10H3;2*1-5H,(H,7,8,9)/q+2;;/p-2

InChI key

XXZSQOVSEBAPGS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atracurium for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nicotinic acetylcholine receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad R Ghodraty et al.
Journal of anesthesia, 26(6), 858-863 (2012-07-04)
During the induction of anesthesia, patients are at risk of aspiration while awaiting full muscle relaxation. Magnesium has been shown to have synergistic effects with neuromuscular blocking drugs. We tested if magnesium, as an adjunct, increases the speed of onset
Michele Carron et al.
Anesthesiology, 117(2), 309-320 (2012-05-23)
The stress responses from tracheal intubation are potentially dangerous in patients with higher cardiovascular risk, such as obese patients. The primary outcome objective of this study was to test whether, in comparison with the endotracheal tube (ETT), the Proseal™ Laryngeal
A Movafegh et al.
European review for medical and pharmacological sciences, 17(4), 447-450 (2013-03-08)
Non-depolarizing neuromuscular blocking agents (NMB) differ in pharmacokinetic and pharmacodynamic parameters. An anesthesiologist according to these similarities and differences is able to choose the least costly one if the same safety profile and same clinical benefit achieved with the different
You-jing Dong et al.
Contemporary clinical trials, 33(3), 482-485 (2012-01-25)
To explore a better administration way through comparison of the pharmacodynamics of cisatracurium administered by continuous infusion or intermittent bolus injection. Thirty patients (ASAI-II) who had no neuromuscular disease and underwent selective surgery under general anesthesia were randomly divided into
U Linstedt et al.
Acta anaesthesiologica Scandinavica, 56(6), 762-769 (2012-03-13)
Use of a single bolus of a hypnotic together with non-depolarizing muscle relaxants for anaesthesia induction may cause inappropriate light levels of anaesthesia (ILLA). The purpose of this study was to compare the incidence of ILLA during anaesthesia induction using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service