Skip to Content
Merck
All Photos(1)

Key Documents

H0200000

Heparin sodium

BRP, European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

heparin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the Issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Heparin sodium EP reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia. Their suitability for any other use is not guaranteed and is the sole responsibility of the user. This standard is not intended for human or animal use.
Established for use in the:

  • Assay of unfractionated heparin according to the Ph. Eur. General text 2.7.5. with assigned contents of 985 IU/mL for anti-IIa and 995 IU/mL for anti-Xa activities
  • Test for heparin in human antithrombin III according to the Ph. Eur. Monograph 0878 with an assigned content of 1035 IU/mL for sheep plasma clotting assay
  • Assay of protamine sulfate according to the prescriptions of the Ph. Eur. Monograph 0569

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assay of Heparin
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 2201-2202 null
Protamine Sulfate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 6947-6949 (2022)
Human Antithrombin III Concentrate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 2835-2837 (2012)
Vanja Cnops et al.
Journal of biomedical materials research. Part A, 108(12), 2473-2483 (2020-05-18)
Neurons of the central nervous system do not regenerate spontaneously after injury. As such, biofunctional tissue scaffolds have been explored to provide a growth-promoting environment to enhance neural regeneration. In this regard, aligned electrospun fibers have proven invaluable for regeneration
Johannes Gojo et al.
Cancer cell, 38(1), 44-59 (2020-07-15)
Ependymoma is a heterogeneous entity of central nervous system tumors with well-established molecular groups. Here, we apply single-cell RNA sequencing to analyze ependymomas across molecular groups and anatomic locations to investigate their intratumoral heterogeneity and developmental origins. Ependymomas are composed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service