Skip to Content
Merck
All Photos(1)

Key Documents

PHR1562

Supelco

Trichloroethylene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Trichloroethylene, TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1601827

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

770 °F

packaging

ampule of 3x1.2 mL each

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Trichloroethylene (TCE) is a chlorine containing organic compound, widely employed as an industrial solvent. It is formed as a major intermediate during the biodegradation of tetrachloroethylene (PCE) in a small anaerobic continuous-flow fixed film column.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4478 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lawrence H Lash et al.
International journal of molecular sciences, 15(11), 21348-21365 (2014-11-21)
Primary cultures of human proximal tubular (hPT) cells are a useful experimental model to study transport, metabolism, cytotoxicity, and effects on gene expression of a diverse array of drugs and environmental chemicals because they are derived directly from the in
Michael A Boles et al.
Journal of the American Chemical Society, 137(13), 4494-4502 (2015-03-17)
This work analyzes the role of hydrocarbon ligands in the self-assembly of nanocrystal (NC) superlattices. Typical NCs, composed of an inorganic core of radius R and a layer of capping ligands with length L, can be described as soft spheres
Xinwei Mao et al.
Applied and environmental microbiology, 81(6), 2015-2024 (2015-01-13)
Dehalococcoides mccartyi 195 (strain 195) and Syntrophomonas wolfei were grown in a sustainable syntrophic coculture using butyrate as an electron donor and carbon source and trichloroethene (TCE) as an electron acceptor. The maximum dechlorination rate (9.9 ± 0.1 μmol day(-1))
Luke Klosterman et al.
Langmuir : the ACS journal of surfaces and colloids, 31(11), 3451-3458 (2015-03-06)
Dopamine-melanin (DM or "polydopamine") can be deposited on virtually any substrate from solution through autoxidation of dopamine. The versatility of this process has allowed surface-mediated assembly of DM for a wide variety of functional coatings. Here we report the impact
Yan Ma et al.
Chemosphere, 121, 117-123 (2014-12-01)
Mechanical soil aeration has long been regarded as an effective ex-situ remediation technique and as suitable for remediation of large-scale sites contaminated by volatile organic compounds (VOCs) at low cost. However, it has been reported that the removal efficiency of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service