Skip to Content
Merck
All Photos(2)

Key Documents

1.00958

Supelco

Trichloroethylene

EMPLURA®

Synonym(s):

Ethylene trichloride, 1,1,2-Trichloroethene, TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
MDL number:
UNSPSC Code:
12191502
EC Index Number:
201-167-4
bp:
86.7 °C (lit.)
vapor pressure:
61 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

4.5 (vs air)

Quality Level

vapor pressure

61 mmHg ( 20 °C)

product line

EMPLURA®

form

liquid

autoignition temp.

770 °F

potency

4920 mg/kg LD50, oral (Rat)
>29000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.9-99 % (v/v) Saturation - at high volume fractions, explosion turns into a decomposition reaction)

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

solubility

1.28 g/L

density

1.463 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service