Merck
  • Many-body effects in nanocrystal superlattices: departure from sphere packing explains stability of binary phases.
Many-body effects in nanocrystal superlattices: departure from sphere packing explains stability of binary phases.
Journal of the American Chemical Society (2015-03-17)
Michael A Boles, Dmitri V Talapin
ABSTRACT
This work analyzes the role of hydrocarbon ligands in the self-assembly of nanocrystal (NC) superlattices. Typical NCs, composed of an inorganic core of radius R and a layer of capping ligands with length L, can be described as soft spheres with softness parameter L/R. Using particle tracking measurements of transmission electron microscopy images, we find that close-packed NCs, like their hard-sphere counterparts, fill space at approximately 74% density independent of softness. We uncover deformability of the ligand capping layer that leads to variable effective NC size in response to the coordination environment. This effect plays an important role in the packing of particles in binary nanocrystal superlattices (BNSLs). Measurements on BNSLs composed of NCs of varying softness in several coordination geometries indicate that NCs deform to produce dense BNSLs that would otherwise be low-density arrangements if the particles remained spherical. Consequently, rationalizing the mixing of two NC species during BNSL self-assembly need not employ complex energetic interactions. We summarize our analysis in a set of packing rules. These findings contribute to a general understanding of entropic effects during crystallization of deformable objects (e.g., nanoparticles, micelles, globular proteins) that can adapt their shape to the local coordination environment.
Sigma-Aldrich
Trichloroethylene, JIS special grade, ≥99.5%
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Supelco
Trichloroethylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Oleic acid, Selectophore, ≥99%