Skip to Content
Merck
All Photos(8)

Key Documents

I8377

Sigma-Aldrich

Iodonitrotetrazolium chloride

Used in colorimetric assays.

Synonym(s):

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, p-Iodonitrotetrazolium Violet, INT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13ClIN5O2
CAS Number:
Molecular Weight:
505.70
Beilstein:
4093224
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥98% (TLC)

form

powder

mp

240 °C (dec.) (lit.)

solubility

H2O: 4 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Cl-].[O-][N+](=O)c1ccc(cc1)-[n+]2nc(nn2-c3ccc(I)cc3)-c4ccccc4

InChI

1S/C19H13IN5O2.ClH/c20-15-6-8-16(9-7-15)23-21-19(14-4-2-1-3-5-14)22-24(23)17-10-12-18(13-11-17)25(26)27;/h1-13H;1H/q+1;/p-1

InChI key

JORABGDXCIBAFL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Electron acceptor for the colorimetric assay of various dehydrogenases
Iodonitrotetrazolium chloride has been used for the staining of cells. It has been used as a coupling agent in the enzymatic assay of D-arabinitol.

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constitutive activation of pp125fak in newly isolated human breast cancer cell lines.
Ignatoski KMW and Ethier SP
Breast Cancer Research and Treatment, 54, 173-173 (1999)
F Mermillod-Blondin et al.
Water research, 39(9), 1687-1698 (2005-05-19)
Biogeochemical processes mediated by microorganisms in river sediments (hyporheic sediments) play a key role in river metabolism. Because biogeochemical reactions in the hyporheic zone are often limited to the top few decimetres of sediments below the water-sediment interface, slow filtration
T J Mankelow et al.
The Biochemical journal, 358(Pt 2), 315-324 (2001-08-22)
Diethyl pyrocarbonate (DEPC), a histidine-modifying reagent, has been utilized to demonstrate the importance of histidine residues in the functioning of proteins. In previous studies of the NADPH oxidase, histidine residues have been determined to be important in the ability of
Sergio Ravasio et al.
Biochemistry, 41(25), 8120-8133 (2002-06-19)
The properties of the recombinant ferredoxin-dependent glutamate synthase of Synechocystis PCC6803 were determined by means of kinetic and spectroscopic approaches in comparison to those exhibited by the bacterial NADPH-dependent enzyme form. The ferredoxin-dependent enzyme was found to be similar to
Yang Hongwei et al.
Ecotoxicology and environmental safety, 59(2), 249-255 (2004-08-26)
Anaerobic biodegradability of organic compounds can be assessed from the changes in organic compounds, the end products, and the activity of microorganisms. In this study, all three of these variables were considered to integrate assessment of the anaerobic biodegradability of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service